Merck
Shrapnel nanoparticles loading docetaxel inhibit metastasis and growth of breast cancer.
Biomaterials (2015-06-25)
Pengfei Xu, Qingshuo Meng, Huiping Sun, Qi Yin, Haijun Yu, Zhiwen Zhang, Mi Cao, Yingyi Zhang, Yaping Li
ZUSAMMENFASSUNG
Metastasis is one of the major obstacles for the successful therapy of breast cancer. To inhibit the metastasis and growth of breast cancer simultaneously, a new docetaxel (DTX) loaded shrapnel nano delivery system with the reduction- and enzyme-sensitive properties was designed and developed. Firstly, methoxy polyethylene glycol-peptide-vitamin E succinate (PPV), a matrix metalloproteinases (MMPs)-sensitive copolymer, was synthesized by conjugating mPEG and vitamin E succinate (VES) using an enzyme-sensitive peptide. Then, DTX loaded methoxy polyethylene glycol-s-s-vitamin E succinate (PSV) micelles (DPM) @ PPV-based liposomes (DPM@PL) were prepared by the incorporation of DPM into the PPV-based liposomes. DPM@PL showed a shrapnel structure with average particle size 113.3 ± 2.7 nm. The drug loading and encapsulation efficiency of DPM@PL were 1.93% and 99.02%, respectively. An obvious burst release (>90%) of drug was observed in the simulated tumor microenvironment with MMPs and reductive glutathione. The cellular uptake and cytotoxicity of DPM@PL in 4T1 cells were significantly enhanced after the pre-treatment of activated MMP-9. Compared with Taxotere(®), DPM@PL remarkably increased the distribution of DTX in lung and tumor of 4T1 tumor-bearing mice, and inhibited the in situ tumor growth and pulmonary metastasis formation effectively through the enhanced DTX-induced apoptosis and the reduced metastasis-promoting proteins expression. Compared with saline group, the inhibitory rates of DPM@PL against tumor volume and lung metastasis were about 81% and 92%, respectively, and it didn't produce the significant systemic toxicity. As a result, DPM@PL could be a promising nano delivery system for the successful therapy of breast cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Natriumpyruvat, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Natriumpyruvat, ReagentPlus®, ≥99%
Sigma-Aldrich
Hämatoxylin
Sigma-Aldrich
Hämatoxylin, certified by the BSC
Sigma-Aldrich
Natriumpyruvat, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sulforhodamin B Mononatriumsalz, Dye content 75 %
Sigma-Aldrich
Sulforhodamin B Natriumsalz, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumpyruvat, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
D-α-Tocopherolsuccinat, semisynthetic, 1210 IU/g
Sigma-Aldrich
Sulforhodamin B Natriumsalz, Technical grade
Sigma-Aldrich
Natriumpyruvat, BioXtra, ≥99%
Sigma-Aldrich
D-α-Tocopherolsuccinat, BioXtra, ≥98.0% (HPLC)
Sigma-Aldrich
Natriumpyruvat, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.