Merck
  • Sub-minute method for simultaneous determination of aspartame, cyclamate, acesulfame-K and saccharin in food and pharmaceutical samples by capillary zone electrophoresis.
Sub-minute method for simultaneous determination of aspartame, cyclamate, acesulfame-K and saccharin in food and pharmaceutical samples by capillary zone electrophoresis.
Journal of chromatography. A (2015-04-22)
Jacqueline Pereira Vistuba, Maressa Danielli Dolzan, Luciano Vitali, Marcone Augusto Leal de Oliveira, Gustavo Amadeu Micke
ZUSAMMENFASSUNG
This paper reports the development of a sub-minute separation method by capillary zone electrophoresis for the determination of aspartame, cyclamate, acesulfame-K and saccharin in food products and pharmaceutical samples. Separations were performed in a fused uncoated silica capillary with UV detection at 220nm. Samples and standards were injected hydrodynamically using the short-end injection procedure. The electrophoretic system was operated under constant voltage of -30kV. The background electrolyte was composed of 45mmolL(-1) 2-amino-2-(hydroxymethyl)-1,3-propanediol and 15mmolL(-1) benzoic acid at pH 8.4. The separation time for all analytes was less than 1min. Evaluation of analytical parameters of the method showed good linearity (r(2)>0.9972), limit of detection of 3.3-6.4mgL(-1), intermediate precision better than 9.75% (peak area of sample) and recovery in the range of 91-117%.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trizma® Base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® Base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Tris(hydroxymethyl)aminomethan, ACS reagent, ≥99.8%
Sigma-Aldrich
Tromethamin, meets USP testing specifications
Sigma-Aldrich
Benzoesäure, ACS reagent, ≥99.5%
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), powder or crystals
Sigma-Aldrich
Trizma® Base, BioUltra, Molecular Biology, ≥99.8% (T)
Sigma-Aldrich
Trizma® Base, ≥99.0% (T)
Sigma-Aldrich
Trizma® Base, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Trizma® Base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® Base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Sigma-Aldrich
Natriumhydroxid, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Natriumhydroxid, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Benzoesäure, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
Sigma-Aldrich
Trizma® Base, 99.7% (T), puriss. p.a.
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Supelco
Acesulfam K, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzoesäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoesäure, ReagentPlus®, 99%
Sigma-Aldrich
Benzoesäure, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
SAFC
Tromethamin
Sigma-Aldrich
Benzoesäure, natural, ≥99.5%, FCC, FG
SAFC
MOPS
Sigma-Aldrich
Benzoesäure, purified by sublimation, ≥99%
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
3-Ethyl-2,4-pentandion, Tautormermischung, 98%