Merck
  • Diverse Targets of β-Catenin during the Epithelial-Mesenchymal Transition Define Cancer Stem Cells and Predict Disease Relapse.
Diverse Targets of β-Catenin during the Epithelial-Mesenchymal Transition Define Cancer Stem Cells and Predict Disease Relapse.
Cancer research (2015-07-01)
Yi-Wen Chang, Ying-Jhen Su, Michael Hsiao, Kuo-Chen Wei, Wei-Hsin Lin, Chi-Lung Liang, Shin-Cheh Chen, Jia-Lin Lee
ZUSAMMENFASSUNG
Wnt signaling contributes to the reprogramming and maintenance of cancer stem cell (CSC) states that are activated by epithelial-mesenchymal transition (EMT). However, the mechanistic relationship between EMT and the Wnt pathway in CSC is not entirely clear. Chromatin immunoprecipitation with high-throughput sequencing (ChIP-seq) indicated that EMT induces a switch from the β-catenin/E-cadherin/Sox15 complex to the β-catenin/Twist1/TCF4 complex, the latter of which then binds to CSC-related gene promoters. Tandem coimmunoprecipitation and re-ChIP experiments with epithelial-type cells further revealed that Sox15 associates with the β-catenin/E-cadherin complex, which then binds to the proximal promoter region of CASP3. Through this mechanism, Twist1 cleavage is triggered to regulate a β-catenin-elicited promotion of the CSC phenotype. During EMT, we documented that Twist1 binding to β-catenin enhanced the transcriptional activity of the β-catenin/TCF4 complex, including by binding to the proximal promoter region of ABCG2, a CSC marker. In terms of clinical application, our definition of a five-gene CSC signature (nuclear β-catenin(High)/nuclear Twist1(High)/E-cadherin(Low)/Sox15(Low)/CD133(High)) may provide a useful prognostic marker for human lung cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-β-Catenin antibody produced in rabbit, whole antiserum