Merck
Deoxycholic acid-grafted PEGylated chitosan micelles for the delivery of mitomycin C.
Drug development and industrial pharmacy (2014-05-03)
Xiu-Rong Zhang, Nian-Qiu Shi, Yang Zhao, He-Yun Zhu, Jiao Guan, Ying Jin
ZUSAMMENFASSUNG
Mitomycin C (MTC) was incorporated to a micelle system preparing from a polymer named deoxycholic acid chitosan-grafted poly(ethylene glycol) methyl ether (mPEG-CS-DA). mPEG-CS-DA was synthesized and characterized by (1)H nuclear magnetic resonance ((1)H-NMR) and Fourier transform infrared spectroscopy. mPEG-CS-DA formed a core-shell micellar structure with a critical micelle concentration of 6.57 µg/mL. The mPEG-CS-DA micelles were spherical with a hydrodynamic diameter of about 231 nm. After poly(ethylene glycol)ylation of deoxycholic acid chitosan (CS-DA), the encapsulation efficiency and drug loading efficiency increased from 50.62% to 56.42% and from 20.51% to 24.13%, respectively. The mPEG-CS-DA micelles possessed a higher drug release rate than the CS-DA micelles. For pharmacokinetics, the area under the curve (AUC) of the mPEG-CS-DA micelles was 1.5 times higher than that of MTC injection, and these micelles can enhance the bioavailability of MTC. mPEG-CS-DA micelles reduced the distribution of MTC in almost all normal tissues and had the potential to improve the kidney toxicity caused by MTC injection.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Deuteriumoxid, 99.9 atom % D
Sigma-Aldrich
Deuteriumoxid, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuteriumoxid, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Tetramethylsilan, ≥99.0% (GC)
Sigma-Aldrich
Deuteriumoxid, filtered, 99.8 atom % D
Sigma-Aldrich
Deuteriumoxid, 99.8 atom % D
Sigma-Aldrich
Deuteriumoxid, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
Deuteriumoxid, 70 atom % D
Sigma-Aldrich
Deuteriumoxid, 60 atom % D
Sigma-Aldrich
Tetramethylsilan, electronic grade, ≥99.99% trace metals basis
Sigma-Aldrich
Deuteriumoxid, 99.9 atom % D