Merck
  • Bioactive, nanostructured Si-substituted hydroxyapatite coatings on titanium prepared by pulsed laser deposition.
Bioactive, nanostructured Si-substituted hydroxyapatite coatings on titanium prepared by pulsed laser deposition.
Journal of biomedical materials research. Part B, Applied biomaterials (2015-01-06)
Julietta V Rau, Ilaria Cacciotti, Sara Laureti, Marco Fosca, Gaspare Varvaro, Alessandro Latini
ZUSAMMENFASSUNG
The aim of this work was to deposit silicon-substituted hydroxyapatite (Si-HAp) coatings on titanium for biomedical applications, since it is known that Si-HAp is able to promote osteoblastic cells activity, resulting in the enhanced bone ingrowth. Pulsed laser deposition (PLD) method was used for coatings preparation. For depositions, Si-HAp targets (1.4 wt % of Si), made up from nanopowders synthesized by wet method, were used. Microstructural and mechanical properties of the produced coatings, as a function of substrate temperature, were investigated by scanning electron and atomic force microscopies, X-ray diffraction, Fourier transform infrared spectroscopy, and Vickers microhardness. In the temperature range of 400-600°C, 1.4-1.5 µm thick Si-HAp films, presenting composition similar to that of the used target, were deposited. The prepared coatings were dense, crystalline, and nanostructured, characterized by nanotopography of surface and enhanced hardness. Whereas the substrate temperature of 750°C was too high and led to the HAp decomposition. Moreover, the bioactivity of coatings was evaluated by in vitro tests in an osteoblastic/osteoclastic culture medium (α-Modified Eagle's Medium). The prepared bioactive Si-HAp coatings could be considered for applications in orthopedics and dentistry to improve the osteointegration of bone implants.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phosphorsäure, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
D-(+)-Glukose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glukose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Phosphorsäure, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Tetraethylorthosilikat, reagent grade, 98%
Sigma-Aldrich
Phosphorsäure, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Calciumhydroxid, ACS reagent, ≥95.0%
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Tetraethylorthosilikat, ≥99.0% (GC)
Sigma-Aldrich
D-(+)-Glukose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Phosphorsäure, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Hydroxylapatit, nanopowder, <200 nm particle size (BET), ≥97%, synthetic
Sigma-Aldrich
D-(+)-Glukose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glukose, ACS reagent
Supelco
D-(+)-Glukose, analytical standard
Sigma-Aldrich
Phosphorsäure -Lösung, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Tetraethylorthosilikat, 99.999% trace metals basis
Sigma-Aldrich
Phosphorsäure, BioUltra, ≥85% (T)
Sigma-Aldrich
Hydroxyapatit, powder, synthetic
Sigma-Aldrich
D-(+)-Glukose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
Phosphorsäure, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphorsäure, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Calciumhydroxid, 99.995% trace metals basis
Sigma-Aldrich
Calciumphosphat tribasisch, 34.0-40.0% Ca basis
Sigma-Aldrich
Hydroxyapatit, synthetic, 99.8% trace metals basis (excludes Mg)
Sigma-Aldrich
D-(+)-Glukose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
Hydroxylapatit, puriss., meets analytical specification of Ph. Eur., BP, FCC, E341, ≥90% (calculated on glowed substance)
Sigma-Aldrich
Hydroxylapatit, nanoparticles, dispersion, 10 wt. % in H2O, <200 nm particle size (BET)
Sigma-Aldrich
Hydroxylapatit, nanopowder, <200 nm particle size (BET), contains 5 wt. % silica as dopant, synthetic
Sigma-Aldrich
Tetraethylorthosilicat, packaged for use in deposition systems