Merck
miR-526a regulates apoptotic cell growth in human carcinoma cells.
Molecular and cellular biochemistry (2015-05-24)
Xiaoli Yang, Cui Wang, Changzhi Xu, Zhifeng Yan, Congwen Wei, Kai Guan, Shengli Ma, Ye Cao, Liping Liu, Deyong Zou, Xiang He, Buchang Zhang, Qingjun Ma, Zirui Zheng
ZUSAMMENFASSUNG
MicroRNAs (miRNAs) play vital roles in the regulation of cell cycle, cell growth, apoptosis, and tumorigenesis. Our previous studies showed that miR-526a positively regulated innate immune response by suppressing CYLD expression, however, the functional relevance of miR-526a expression and cell growth remains to be evaluated. In this study, miR-526a overexpression was found to promote cancer cell proliferation, migration, and anchor-independent colony formation. The molecular mechanism(s) of miR-526a-mediated growth stimulation is associated with rapid cell cycle progression and inhibition of cell apoptosis by targeting CYLD. Taken together, these results provide evidence to show the stimulatory role of miR-526a in tumor migration and invasion through modulation of the canonical NF-κB signaling pathway.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Glycerin, FCC, FG
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Natriumdodecylsulfat, ≥90% ((Assay))
Sigma-Aldrich
Natriumdodecylsulfat, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
L-(−)-Dithiothreitol, ≥95% (titration)
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri