Merck
  • Detecting and Number Counting of Single Engineered Nanoparticles by Digital Particle Polymerase Chain Reaction.
Detecting and Number Counting of Single Engineered Nanoparticles by Digital Particle Polymerase Chain Reaction.
ACS nano (2015-08-11)
Daniela Paunescu, Carlos A Mora, Lorenzo Querci, Reinhard Heckel, Michela Puddu, Bodo Hattendorf, Detlef Günther, Robert N Grass
ZUSAMMENFASSUNG
The concentrations of nanoparticles present in colloidal dispersions are usually measured and given in mass concentration (e.g. mg/mL), and number concentrations can only be obtained by making assumptions about nanoparticle size and morphology. Additionally traditional nanoparticle concentration measures are not very sensitive, and only the presence/absence of millions/billions of particles occurring together can be obtained. Here, we describe a method, which not only intrinsically results in number concentrations, but is also sensitive enough to count individual nanoparticles, one by one. To make this possible, the sensitivity of the polymerase chain reaction (PCR) was combined with a binary (=0/1, yes/no) measurement arrangement, binomial statistics and DNA comprising monodisperse silica nanoparticles. With this method, individual tagged particles in the range of 60-250 nm could be detected and counted in drinking water in absolute number, utilizing a standard qPCR device within 1.5 h of measurement time. For comparison, the method was validated with single particle inductively coupled plasma mass spectrometry (sp-ICPMS).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Tetraethylorthosilikat, reagent grade, 98%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Essigsäure -Lösung, suitable for HPLC
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Tetraethylorthosilikat, ≥99.0% (GC)
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Tetraethylorthosilikat, 99.999% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Sigma-Aldrich
Essigsäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Essigsäure, natural, ≥99.5%, FG
Sigma-Aldrich
3-(Trimethoxysilyl)-propyl-N,N,N-trimethylammonium-15N-chlorid, 99 atom % 15N, 97% (CP)
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, purum, ≥99.0% (TLC)
Sigma-Aldrich
Tetraethylorthosilicat, packaged for use in deposition systems
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Essigsäure-12C2, 99.9 atom % 12C