Merck
  • Mass-spectrometry-based method for screening of new peptide ligands for G-protein-coupled receptors.
Mass-spectrometry-based method for screening of new peptide ligands for G-protein-coupled receptors.
Analytical and bioanalytical chemistry (2015-05-04)
Camila T Cologna, Nicolas Gilles, Julien Echterbille, Michel Degueldre, Denis Servent, Edwin de Pauw, Loïc Quinton
ZUSAMMENFASSUNG
G-protein-coupled receptors (GPCRs) constitute the largest family of transmembrane proteins. Although implicated in almost all physiological processes in the human body, most of them remain unexploited, mostly because of the lack of specific ligands. The objective of this work is to develop a new mass-spectrometry-based technique capable of identifying new peptide ligands for GPCRs. The strategy is based on the incubation of cellular membranes overexpressing GPCRs with a mixture of peptides that contains potential ligands. Peptide ligands bind to the receptors, whereas other peptides remain in the binding buffer. Bound peptides are eluted from membranes and directly detected, identified, and characterized by MALDI TOF-TOF. The results reveal the efficacy of the procedure for selecting a specific ligand of GPCRs in both simple and complex mixtures of peptides. This new approach may offer direct purification, identification, and characterization of the new ligand in a single workflow. The proposed method is labeling-free and, unlike radio-binding and other techniques, it does not require a previously known labeled ligand of the studied GPCR. All these properties greatly reduce the experimental constraints. Moreover, because it is not based on the principle of a competitive specific binding, this technique constitutes a new tool to discover new ligands not only for known GPCRs, but also for orphan GPCRs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Salzsäure -Lösung, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Salzsäure -Lösung, 32 wt. % in H2O, FCC