Merck
  • Airway inflammation and ammonia exposure among female Palestinian hairdressers: a cross-sectional study.
Airway inflammation and ammonia exposure among female Palestinian hairdressers: a cross-sectional study.
Occupational and environmental medicine (2015-02-06)
Maysaa Nemer, Liv I B Sikkeland, Mayes Kasem, Petter Kristensen, Khaldoun Nijem, Espen Bjertness, Øivind Skare, Berit Bakke, Johny Kongerud, Marit Skogstad
ZUSAMMENFASSUNG
Little is known about the working conditions and airway inflammation in hairdressers in Palestine. We aimed to investigate if hairdressers in Palestine have a higher level of airway inflammation as compared to a control group. We also assessed the hairdressers' physical working conditions and exposure to ammonia gases at the hair salons. Lastly, we investigated the association between ammonia levels and inflammation markers in the airways and the blood. Our study participants were 33 non-smoking hairdressers (aged 19-50 years) and 35 non-smoking control subjects (aged 18-49 years). Both groups answered a questionnaire on respiratory symptoms, and performed lung function and exhaled nitric oxide (eNO) tests. Blood and sputum samples were collected from all participants and air concentration levels of ammonia were measured in 13 salons. Hairdressers had a higher level of sputum neutrophil count (absolute numbers/mg sputum (median (25th-75th centiles)) compared to controls, 376 (183-980) and 182 (96-358), respectively. Hairdressers also had significantly elevated eNO and blood C reactive protein (CRP) levels compared to the control subjects, controlled for age and body mass index. Exposure measurements showed that the hairdressers in salons with scarce ventilation were exposed to ammonia concentration, ranging from 3 to 61 mg/m(3). Compared to unexposed controls, the hairdressers had signs of neutrophilic airway inflammation, higher eNO levels and higher CRP. The hairdressers were exposed to high concentrations of ammonia from hairdressing chemicals and their working conditions were unsatisfactory.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trypanblau -Lösung, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Trypanblau, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Trypanblau, ≥80% (HPLC), Dye content 60 %