Merck
  • Synthesis and in vitro kinetic evaluation of N-thiazolylacetamido monoquaternary pyridinium oximes as reactivators of sarin, O-ethylsarin and VX inhibited human acetylcholinesterase (hAChE).
Synthesis and in vitro kinetic evaluation of N-thiazolylacetamido monoquaternary pyridinium oximes as reactivators of sarin, O-ethylsarin and VX inhibited human acetylcholinesterase (hAChE).
Bioorganic & medicinal chemistry (2015-06-06)
Aditya Kapil Valiveti, Uma M Bhalerao, Jyotiranjan Acharya, Hitendra N Karade, Badri Narayan Acharya, G Raviraju, Anand K Halve, Mahabir Parshad Kaushik
ZUSAMMENFASSUNG
Presently available medications for treatment of organiphosphorus poisoning are not sufficiently effective due to various pharmacological and toxicological reasons. In this regard, herein we report the synthesis of a series of N-thiazolylacetamide monoquaternary pyridinium oximes and its analogs (1a-1b to 6a-6b) with diversely substituted thiazole ring and evaluation of their in vitro reactivation efficacies against nerve agent (sarin, O-ethylsarin and VX) inhibited human erythrocyte acetylcholinesterase (hAChE). Reactivation kinetics was performed to determine dissociation constant (KD), reactivity rate constant (kr) and the second order rate constant (kr2) for all the compounds and compared their efficacies with commercial antidotes viz. 2-PAM and obidoxime. All the newly synthesized oximes were evaluated for their physicochemical parameters (pKa) and correlated with their respective reactivation efficacies to assess the capability of the oxime reactivator. Three of these novel compounds showed promising reactivation efficacies toward OP inhibited hAChE. Molecular docking studies were performed in order to correlate the reactivation efficacies with their interactions in the active site of the AChE.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid-d6, 99.9 atom % D
Sigma-Aldrich
Glycin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Glycin, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Kaliumphosphat, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethylsulfoxid-d6, 99.5 atom % D
Sigma-Aldrich
Glycin, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Glycin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Isopropylalkohol, ≥99.7%, FCC, FG
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Kaliumphosphat, ReagentPlus®
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Kaliumphosphat, Molecular Biology, ≥98.0%
Sigma-Aldrich
Kaliumphosphat, anhydrous, suitable for luminescence, Molecular Biology, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Salzsäure -Lösung, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Dimethylsulfoxid-d6, "100%", 99.96 atom % D
SAFC
Glycin
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Kaliumphosphat, reagent grade, ≥98.0%
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Dimethylsulfoxid-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Dimethylsulfoxid-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Kaliumphosphat, BioUltra, Molecular Biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate dibasic, meets USP testing specifications
Sigma-Aldrich
Kaliumphosphat -Lösung, 1.0 M