Merck
  • Starch/silver nanocomposite: Effect of thermal treatment temperature on the morphology, oxygen and water transport properties.
Starch/silver nanocomposite: Effect of thermal treatment temperature on the morphology, oxygen and water transport properties.
Carbohydrate polymers (2015-10-03)
Perrine Cheviron, Fabrice Gouanvé, Eliane Espuche
ZUSAMMENFASSUNG
The present work reports a strategy involving the preparation of nanostructured starch based film containing silver nanoparticles (AgNPs) using a completely green chemistry process. The nanocomposite films were prepared by solution cast process. The AgNPs were in situ generated inside the polymer film by thermal treatment at different temperatures (25, 40 and 85 °C). The influence of the presence and the amount of reducing agent (glucose) were also investigated. For all nanocomposite films, the AgNPs were spherical with a diameter less than 15 nm. Contrary to the presence of glucose, thermal treatment condition was a key factor for the AgNPs structure. Crystalline AgNPs were obtained only after thermal treatment at 85 °C. Improvements of water and oxygen barrier properties near to one decade were observed in this last case and were explained by the formation of crystalline AgNPs associated to the establishment of strong interactions between AgNPs and starch polymer matrix.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Sigma-Aldrich
Silbernitrat, ACS reagent, ≥99.0%
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Silbernitrat, ReagentPlus®, ≥99.0% (titration)
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Silbernitrat, 99.9999% trace metals basis
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerin, FCC, FG
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Silbernitrat auf Kieselgel, extent of labeling: ~10 wt. % loading, +230 mesh
Sigma-Aldrich
Silbernitrat -Lösung, 2.5 % (w/v) AgNO3 in H2O
Sigma-Aldrich
Silbernitrat, BioReagent, suitable for plant cell culture, >99% (titration)
Sigma-Aldrich
Silbernitrat, ≥99.999% trace metals basis
Sigma-Aldrich
Silbernitrat, BioXtra, >99% (titration)