Merck
  • Evaluation of Chicken IgY Generated Against Canine Parvovirus Viral-Like Particles and Development of Enzyme-Linked Immunosorbent Assay and Immunochromatographic Assay for Canine Parvovirus Detection.
Evaluation of Chicken IgY Generated Against Canine Parvovirus Viral-Like Particles and Development of Enzyme-Linked Immunosorbent Assay and Immunochromatographic Assay for Canine Parvovirus Detection.
Viral immunology (2015-10-16)
Jinxin He, Yuan Wang, Shiqi Sun, Xiaoying Zhang
ZUSAMMENFASSUNG
Immunoglobulin Y (IgY) antibodies were generated against canine parvovirus virus-like particles (CPV-VLPs) antigen using chickens. Anti-CPV-VLPs-IgY was extracted from hen egg yolk and used for developing enzyme-linked immunosorbent assay (ELISA) and immunochromatographic assay (ICA) for the detection of CPV in dog feces. The cutoff negative values for anti-CPV-VLPs-IgY were determined using negative fecal samples (already confirmed by polymerase chain reaction [PCR]). In both ELISA and ICA, there was no cross-reaction with other diarrheal pathogens. Thirty-four fecal samples were collected from dogs with diarrhea, of which 26.47% were confirmed as CPV-positive samples by PCR, while 29.41% and 32.35% of the samples were found to be positive by ELISA and ICA, respectively. The developed ELISA and ICA exhibited 97.06% and 94.12% conformity with PCR. Higher sensitivity and specificity were observed for IgY-based ELISA and ICA. Thus, they could be suitable for routine use in the diagnosis of CPV in dogs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Isocyanatethylmethacrylat, contains ≤0.1% BHT as inhibitor, 98%