Merck
  • Differentiation of rye and wheat flour as well as mixtures by using the kinetics of Karl Fischer water titration.
Differentiation of rye and wheat flour as well as mixtures by using the kinetics of Karl Fischer water titration.
Food chemistry (2015-11-18)
Daniel I Hădărugă, Corina I Costescu, Laura Corpaş, Nicoleta G Hădărugă, Heinz-Dieter Isengard
ZUSAMMENFASSUNG
The aim of this study was to find a simple way to differentiate between rye and wheat flour and their mixtures by using the kinetic parameters of Karl Fischer water titration (KFT). Consequently, the water content and type of molecules in rye and wheat mixtures used in Romanian bread making have been investigated by means of volumetric KFT. Further, the kinetics of KFT have been determined and novel kinetic parameters corresponding to "surface" and "strongly-retained" water molecules have been identified to discriminate between rye and wheat flour and their mixtures. The "surface" and "strongly-retained" water reaction rates well correlate with the rye content in the flour mixtures, especially at higher temperature analysis (r>0.95). These parameters can be used as indicators for quality evaluation of such type of mixtures, as well as to identify adulteration by improper use of the rye-wheat flour ratios in bread making.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Imidazol, ReagentPlus®, 99%
Sigma-Aldrich
Imidazol, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Imidazol, Molecular Biology, ≥99% (titration)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Imidazol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Imidazol-Pufferlösung, BioUltra, 1 M in H2O
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Imidazol, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Schwefeldioxid, ≥99.9%
Sigma-Aldrich
Kaliumfluorid, powder, ≥99.9% trace metals basis
Sigma-Aldrich
Kaliumfluorid, ≥99.97% trace metals basis
Sigma-Aldrich
Imidazol, ≥99% (titration), crystalline
Sigma-Aldrich
Kaliumfluorid, BioUltra, ≥99.5% (F)
Sigma-Aldrich
Imidazol, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Imidazol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Imidazol, ReagentPlus®, 99%, Redi-Dri, free-flowing
Sigma-Aldrich
Imidazol, Molecular Biology, ≥99% (titration), free-flowing, Redi-Dri