Merck
  • Comparing the electronic relaxation dynamics of aniline and d(7)-aniline following excitation at 272-238 nm.
Comparing the electronic relaxation dynamics of aniline and d(7)-aniline following excitation at 272-238 nm.
Physical chemistry chemical physics : PCCP (2015-05-21)
Oliver M Kirkby, Matthieu Sala, Garikoitz Balerdi, Rebeca de Nalda, Luis Bañares, Stéphane Guérin, Helen H Fielding
ZUSAMMENFASSUNG
Femtosecond time-resolved photoelectron spectroscopy experiments have been used to compare the electronic relaxation dynamics of aniline and d7-aniline following photoexcitation in the range 272-238 nm. Together with the results of recent theoretical investigations of the potential energy landscape [M. Sala, O. M. Kirkby, S. Guérin and H. H. Fielding, Phys. Chem. Chem. Phys., 2014, 16, 3122], these experiments allow us to resolve a number of unanswered questions surrounding the nonradiative relaxation mechanism. We find that tunnelling does not play a role in the electronic relaxation dynamics, which is surprising given that tunnelling plays an important role in the electronic relaxation of isoelectronic phenol and in pyrrole. We confirm the existence of two time constants associated with dynamics on the 1(1)πσ* surface that we attribute to relaxation through a conical intersection between the 1(1)πσ* and 1(1)ππ* states and motion on the 1(1)πσ* surface. We also present what we believe is the first report of an experimental signature of a 3-state conical intersection involving the 2(1)ππ*, 1(1)πσ* and 1(1)ππ* states.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anilin, ACS reagent, ≥99.5%
Sigma-Aldrich
Anilin, ReagentPlus®, 99%