Merck
A dendrimer-like hyper branched chitosan beads toward fluoride adsorption from water.
International journal of biological macromolecules (2015-04-12)
Subbaiah Muthu Prabhu, Sankaran Meenakshi
ZUSAMMENFASSUNG
The present study reports a novel approach for the preparation of polyamidoamine grafted chitosan beads (PAAGCB) via Michael addition followed by protonation of PAAGCB to get protonated PAAGCB (H(+)-PAAGCB). Various metal ions viz., Al(3+), Ce(3+), La(3+) and Zr(4+) have been loaded onto the PAAGCB in order to get respective Al-PAAGCB, Ce-PAAGCB, La-PAAGCB and Zr-PAAGCB. All these sorbents were used to remove the fluoride ions from aqueous solution. Ethylenediamine is used for amination purpose of chitosan beads through Michael addition. The results showed that Zr-PAAGCB was more selective with the maximum defluoridation capacity of 17.47 mg/g than the other metal ion loaded chitosan beads. The sorbents were characterized using FTIR, XRD, SEM and EDAX. The fluoride sorption was reasonably explained using Freundlich and Langmuir isotherm models. The mechanism concerned in the adsorption of fluoride ions is by physisorption on heterogeneous materials according to the results obtained by fitting the data to various isotherm models. Temperature study carried out at 303, 313 and 323K revealed that the adsorption process was spontaneous and endothermic in nature. The applicability of the sorbents studied has been tested with field sample collected from fluoride endemic area.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Ethylendiamin, ReagentPlus®, ≥99%
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyd -Lösung, 50 wt. % in H2O
Sigma-Aldrich
Aluminiumoxid, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Methylacrylat, 99%, contains ≤100 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Sigma-Aldrich
Aluminiumoxid, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Aluminiumsulfat Hydrat, 99.99% trace metals basis
Sigma-Aldrich
Ethylendiamin, purified by redistillation, ≥99.5%
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Aluminiumoxid, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Natriumfluorid, 99.99% trace metals basis
Sigma-Aldrich
Essigsäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Essigsäure, natural, ≥99.5%, FG
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Lanthan(III)-chlorid, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminiumoxid, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O
Sigma-Aldrich
Aluminiumoxid, 99.997% trace metals basis
Sigma-Aldrich
Natriumfluorid, BioXtra, ≥99%
Sigma-Aldrich
Natriumfluoridlösung
Sigma-Aldrich
Glutarsäure-Dialdehyd -Lösung, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyd -Lösung, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Natriumfluorid, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Lanthan(III)-chlorid, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Aluminiumoxid, single crystal substrate, <0001>
Sigma-Aldrich
Ethylendiamin, BioXtra
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, purum, ≥99.0% (TLC)