Merck
  • Pretreatment effects on the sorption of Cr(VI) onto surfactant-modified zeolite: Mechanism analysis.
Pretreatment effects on the sorption of Cr(VI) onto surfactant-modified zeolite: Mechanism analysis.
Journal of environmental management (2015-08-02)
Wenzhe Song, Taihong Shi, Di Yang, Jinpeng Ye, Yi Zhou, Yuxiang Feng
ZUSAMMENFASSUNG
Adsorption of Cr(VI) onto different pretreated zeolites modified with cetylpyridinium chloride (CPC) is investigated using batch studies, Fourier-transform infrared spectroscopy (FT-IR), X-ray diffraction (XRD) measurements and scanning electron microscopy (SEM). The results indicate that acidification after alkalization is the most effective pretreatment method, and only alkalization would significantly reduce the sorption capability. This behavior is due to the precipitates, such as CaCO3 and MgCO3, generated after alkalization on zeolite surface interfere the formation of the CPC bilayer, which provides active sites for sorbing Cr(VI). The schematic of the adsorption mechanism is presented. The results indicate that developing a better understanding of the influence of different pretreatments is quite helpful and suggest that the content of Ca/Mg/Fe could be a good indication of the effectiveness of preprocessing.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, Molecular Biology, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, BioXtra, ≥99%
Sigma-Aldrich
Cetylpyridiniumchlorid, meets USP testing specifications
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, BioUltra, Molecular Biology, ≥99.0% (AT)