Merck
  • Complete regression of breast tumour with a single dose of docetaxel-entrapped core-cross-linked polymeric micelles.
Complete regression of breast tumour with a single dose of docetaxel-entrapped core-cross-linked polymeric micelles.
Biomaterials (2015-04-22)
Qizhi Hu, Cristianne J Rijcken, Ruchi Bansal, Wim E Hennink, Gert Storm, Jai Prakash
ZUSAMMENFASSUNG
Treatment with chemotherapy such as docetaxel (DTX) is associated with significant toxicity and tumour recurrence. In this study, we developed DTX-entrapped core-cross-linked polymeric micelles (DTX-CCL-PMs, 66 nm size) by covalently conjugating DTX to CCL-PMs via a hydrolysable ester bond. The covalent conjugation allowed for sustained release of DTX under physiological conditions in vitro. In vivo, DTX-CCL-PMs demonstrated superior therapeutic efficacy in mice bearing MDA-MB-231 tumour xenografts as compared to the marketed formulation of DTX (Taxotere(®)). Strikingly, a single intravenous injection of DTX-CCL-PMs enabled complete regression of both small (∼150 mm(3)) and established (∼550 mm(3)) tumours, leading to 100% survival of the animals. These remarkable antitumour effects of DTX-CCL-PMs are attributed to its enhanced tumour accumulation and anti-stromal activity. Furthermore, DTX-CCL-PMs exhibited superior tolerability in healthy rats as compared to Taxotere. These preclinical data strongly support clinical translation of this novel nanomedicinal product for the treatment of cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Triethylamin, ≥99.5%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, BioReagent, suitable for electrophoresis, ≥99.0%
Sigma-Aldrich
4-(Dimethylamino)-pyridin, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Triethylamin, ≥99%
Sigma-Aldrich
N,N′-Dicyclohexylcarbodiimid, 99%
Sigma-Aldrich
Triethylamin, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, BioReagent, Molecular Biology, ≥99% (GC)
Sigma-Aldrich
Triethylamin, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Ammoniumacetat, Molecular Biology, ≥98%
Sigma-Aldrich
Milchsäure, meets USP testing specifications
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, ≥99.5%, purified by redistillation
Sigma-Aldrich
Ammoniumacetat -Lösung, Molecular Biology, 7.5 M
Sigma-Aldrich
Milchsäure, 88%, FCC
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, ReagentPlus®, 99%
Sigma-Aldrich
Milchsäure, natural, ≥85%
Sigma-Aldrich
Ammoniumacetat, 99.999% trace metals basis
Sigma-Aldrich
Milchsäure -Lösung, ACS reagent, ≥85%
Sigma-Aldrich
Kaliumpersulfat, 99.99% trace metals basis
Sigma-Aldrich
2-Chlor-1-methylpyridiniumiodid, 97%
Sigma-Aldrich
Ammoniumacetat, reagent grade, ≥98%
Sigma-Aldrich
Ammoniumacetat, BioXtra, ≥98%
Sigma-Aldrich
Triethylamin, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Ameisensäure, ≥95%, FCC, FG
Sigma-Aldrich
N,N′-Dicyclohexylcarbodiimid -Lösung, 1.0 M in methylene chloride
Sigma-Aldrich
DL-Milchsäure, 85 % (w/w), syrup
Sigma-Aldrich
Triethylamin, ≥99.5%
Sigma-Aldrich
Triethylamin, ampule, ≥99.5% (GC)