Merck
  • Behavioural changes observed in demyelination model shares similarities with white matter abnormalities in humans.
Behavioural changes observed in demyelination model shares similarities with white matter abnormalities in humans.
Behavioural brain research (2015-04-07)
Nathalia Serra-de-Oliveira, Sabine Nunes Boilesen, Carolina Prado de França Carvalho, Luciana LeSueur-Maluf, Ricardo de Lima Zollner, Regina Célia Spadari, Carla Cristina Medalha, Gláucia Monteiro de Castro
ZUSAMMENFASSUNG
Multiple sclerosis (MS) is a chronic, inflammatory, demyelinating disease of the central nervous system (CNS). Further to the symptoms resulting from demyelination, new studies point to the involvement of neuroinflammation and white matter abnormalities in psychiatric disorders and neurodegenerative diseases. Cuprizone, a model of MS, produces consistent demyelination and elicits behavioural, morphological and inflammatory changes in animals that share some similarities with those observed in humans. In this study, we used the cuprizone model in Lewis rats to evaluate clinical signs triggered by the demyelination process which could be comparable with the symptoms seen in white matter abnormalities in human beings. To induce the demyelination process, 0.6% cuprizone was added to the Lewis rats' diet for 4 weeks. We proceeded with behavioural, morphological and immunological analyses. Animals fed with cuprizone exhibited behavioural changes: higher scores in the neurotoxicity test, reduced exploratory and locomotion behaviour, and also an increase of permanency in the closed arm of the elevated plus maze test, were observed. In these analyses, the animals showed motor coordination impairment and anxiety-like behaviour. Demyelination also triggered changes in discrimination of objects identified by an increase in the time spent close to a familiar object. These behavioural alterations were associated with a significant increase in the levels of TNF-alpha and corticosterone, consistent with the activation of microglia and astrocytes. Taken together, the results of this work show the cuprizone/Lewis rat model demyelination as an attractive paradigm for studying the correlation between white matter abnormalities and behaviour.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Bis(cyclohexanon)oxaldihydrazon, ≥95% (TLC)
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Sigma-Aldrich
o-Xylen, anhydrous, 97%
Sigma-Aldrich
Essigsäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Essigsäure, natural, ≥99.5%, FG
Sigma-Aldrich
Lithiumcarbonat, 99.999% trace metals basis
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Lithiumcarbonat, 99.997% trace metals basis
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, purum, ≥99.0% (TLC)
Sigma-Aldrich
Lithium-7Li2-carbonat, ≥99 atom % 7Li, 99% (CP)
Sigma-Aldrich
Essigsäure-12C2, 99.9 atom % 12C