Merck
  • Development and optimization of taste-masked orally disintegrating tablets (ODTs) of clindamycin hydrochloride.
Development and optimization of taste-masked orally disintegrating tablets (ODTs) of clindamycin hydrochloride.
Drug development and industrial pharmacy (2014-07-08)
Stuart L Cantor, Mansoor A Khan, Abhay Gupta
ZUSAMMENFASSUNG
The purpose of this research was to develop an orally disintegrating tablet (ODT) dosage form containing taste-masked beads of clindamycin HCl. Several formulation strategies were evaluated and a taste-masked ODT of clindamycin HCl was prepared without the use of a waxy cushioning agent. Clindamycin HCl (ca. 46% w/w) was coated onto microcrystalline cellulose beads (Cellets® 200) followed by the addition of a taste-masking layer of amino methacrylate copolymer, NF (Eudragit EPO® (EPO)) coating suspension. The efficiency of both the drug coating process and the taste-masking polymer coating process, as well as the taste masking ODTs was determined using potency and drug release analysis. Magnesium stearate was found to be advantageous over talc in improving the efficiency of the EPO coating suspension. A response surface methodology using a Box-Behnken design for the tablets revealed compression force and levels of both disintegrant and talc to be the main factors influencing the ODT properties. Blending of talc to the EPO-coated beads was found to be the most critical factor in ensuring that ODTs disintegrate within 30 s. The optimized ODTs formulation also showed negligible (<0.5%) drug release in 1 min using phosphate buffer, pH 6.8 (which is analogous to the residence time and pH in the oral cavity). By carefully adjusting the levels of coating polymers, the amounts of disintegrant and talc, as well as the compression force, robust ODTs can be obtained to improve pediatric and geriatric patient compliance for clindamycin oral dosage forms.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Stearinsäure, Grade I, ≥98.5% (capillary GC)
Sigma-Aldrich
Stearinsäure, reagent grade, 95%
Sigma-Aldrich
Stearinsäure, ≥95%, FCC, FG
Sigma-Aldrich
Magnesiumstearat, technical grade
Sigma-Aldrich
Magnesiumstearat, puriss., meets analytical specification of Ph. Eur., BP, ≥90% stearic and palmitic acid basis, ≥40% stearic acid basis (GC), 4.0-5.0% Mg basis (calc on dry sub.)