Merck
  • The Zinc Ion Chelating Agent TPEN Attenuates Neuronal Death/apoptosis Caused by Hypoxia/ischemia Via Mediating the Pathophysiological Cascade Including Excitotoxicity, Oxidative Stress, and Inflammation.
The Zinc Ion Chelating Agent TPEN Attenuates Neuronal Death/apoptosis Caused by Hypoxia/ischemia Via Mediating the Pathophysiological Cascade Including Excitotoxicity, Oxidative Stress, and Inflammation.
CNS neuroscience & therapeutics (2015-07-21)
Wei-Ming Wang, Zhao Liu, Ai-Jun Liu, Yu-Xiang Wang, Hong-Gang Wang, Di An, Bin Heng, Lai-Hua Xie, Jun-Li Duan, Yan-Qiang Liu
ZUSAMMENFASSUNG
We aim to determine the significant effect of TPEN, a Zn(2+) chelator, in mediating the pathophysiological cascade in neuron death/apoptosis induced by hypoxia/ischemia. We conducted both in vivo and in vitro experiments in this study. PC12 cells were used to establish hypoxia/ischemia model by applying oxygen-glucose deprivation (OGD). SHR-SP rats were used to establish an acute ischemic model by electrocoagulating middle cerebral artery occlusion. The effect of TPEN on neuron death/apoptosis was evaluated. In addition, the relative biomarks of excitotoxicity, oxidative stress, and inflammation reactions in hypoxia/ischemia PC12 cell model as well as in SHR-SP rat hypoxia/ischemia model were also assessed. TPEN significantly attenuates the neurological deficit, reduced the cerebral infarction area and the ratio of apoptotic neurons, and increased the expression of GluR2 in the rat hypoxia/ischemia brain. TPEN also increased blood SOD activity, decreased blood NOS activity and blood MDA and IL-6 contents in rats under hypoxia/ischemia. In addition, TPEN significantly inhibited the death and apoptosis of cells and attenuated the alteration of GluR2 and NR2 expression caused by OGD or OGD plus high Zn(2+) treatments. Zn(2+) is involved in neural cell apoptosis and/or death caused by hypoxia/ischemia via mediating excitotoxicity, oxidative stress, and inflammation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trizma® Base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Trizma® Base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, BioReagent, suitable for electrophoresis, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Tris(hydroxymethyl)aminomethan, ACS reagent, ≥99.8%
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Tromethamin, meets USP testing specifications
Sigma-Aldrich
Natriumdithionit, ≥82% (RT)
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, BioReagent, Molecular Biology, ≥99% (GC)
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), powder or crystals
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Trizma® Base, BioUltra, Molecular Biology, ≥99.8% (T)
Sigma-Aldrich
Ethylendiamin, ReagentPlus®, ≥99%
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, ≥99.5%, purified by redistillation
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, ReagentPlus®, 99%
Sigma-Aldrich
Trizma® Base, ≥99.0% (T)
Sigma-Aldrich
4,4′-Diaminodiphenylmethan, ≥97.0% (GC)
Sigma-Aldrich
Glycerin, FCC, FG
Sigma-Aldrich
Trizma® Base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)