Merck
  • Influence of elemental impurities in aluminum hydroxide adjuvant on the stability of inactivated Japanese Encephalitis vaccine, IXIARO®.
Influence of elemental impurities in aluminum hydroxide adjuvant on the stability of inactivated Japanese Encephalitis vaccine, IXIARO®.
Vaccine (2015-06-23)
Robert Schlegl, Michael Weber, Jürgen Wruss, Donald Low, Kirsten Queen, Shaun Stilwell, Erik B Lindblad, Michael Möhlen
ZUSAMMENFASSUNG
Aluminum hydroxide is a critical raw material in the production of many vaccines. It is used as an adjuvant in the formulation of the final bulk vaccine, and for this it must meet the specifications of the European Pharmacopeia Monograph. We investigated whether vaccine stability was affected by the presence of trace amounts of elemental impurities in commercially available aluminum hydroxide. The content of residual elemental impurities in commercially available aluminum hydroxide was determined by selective and sensitive inductively coupled-plasma mass spectrometry and inductively coupled plasma atomic emission spectroscopy. We found significant differences between different suppliers, but also between different lots from the same supplier. Inactivated Japanese encephalitis vaccine, IXIARO(®), was used to study the effect of residual metals in aluminum hydroxide on antigen stability. We propose that antigen degradation occurred via a pathway involving the metal-catalyzed, auto-oxidation of a process-related impurity (sulfite). Thus, sulfite auto-oxidation resulted in antigen degradation when residual Cu was present at elevated concentrations in aluminum hydroxide.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyd -Lösung, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
SAFC
Formaldehyd -Lösung, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Natriummetabisulfit, ReagentPlus®, ≥99%
Sigma-Aldrich
Natriummetabisulfit, reagent grade, 97%
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Kupfer(II)-chlorid, 97%
Sigma-Aldrich
Natriummetabisulfit, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E223, dry, 97-100.5%
Sigma-Aldrich
Kupfer(II)-chlorid, powder, 99%
Sigma-Aldrich
Formaldehyd -Lösung, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Kupfer(II)-chlorid, powder, ≥99.995% trace metals basis
Supelco
Formaldehyd -Lösung, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Kupfer(II)-chlorid, 99.999% trace metals basis
Sigma-Aldrich
Formaldehyd-12C -Lösung, 20% in H2O, 99.9 atom % 12C