Merck
  • Structure and aggregation properties of a Schiff-base zinc(II) complex derived from cis-1,2-diaminocyclohexane.
Structure and aggregation properties of a Schiff-base zinc(II) complex derived from cis-1,2-diaminocyclohexane.
Dalton transactions (Cambridge, England : 2003) (2015-06-24)
Giuseppe Consiglio, Ivan Pietro Oliveri, Francesco Punzo, Amber L Thompson, Santo Di Bella, Salvatore Failla
ZUSAMMENFASSUNG
This contribution explores the effect of the bridging diamine upon the aggregation properties of a Zn(II) Schiff-base complex, , both in the solid state and in solution. The X-ray structure of , resulting from the harvest of good quality crystals using chloroform and diethyl ether as solvents, shows the presence of a densely packed dimer in the solid state which pentacoordinates two Zn atoms involved in a μ-phenoxo bridge. Detailed studies in solution, through (1)H NMR, DOSY NMR, and optical spectroscopic investigations, indicate the typical aggregation/deaggregation behaviour on switching from non-coordinating to coordinating solvents, in relation to the Lewis acidic character of such Zn(II) complexes. Thus, while in DMSO-d6 both (1)H NMR and DOSY studies suggest the existence of monomeric species, in chloroform solution experimental data support the existence of aggregates. However, unlike our previous studies, (1)H NMR data in chloroform solution indicate the existence of an asymmetric dimer, as observed in the X-ray crystal structure. This further evidences a very rigid backbone of the dimeric aggregate and can be related to the defined stereochemistry of the chelate cis-1,2-diaminocyclohexane bridge.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Triethylamin, ≥99.5%
Sigma-Aldrich
Triethylamin, ≥99%
Sigma-Aldrich
Triethylamin, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamin, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
2-Methyl-2-buten, ≥95.0% (GC)
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Triethylamin, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Triethylamin, ≥99.5%
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Triethylamin, ampule, ≥99.5% (GC)
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Chloroform-d, 99.8 atom % D