Merck
  • Optimization of ultrasonic assisted continuous production of biodiesel using response surface methodology.
Optimization of ultrasonic assisted continuous production of biodiesel using response surface methodology.
Ultrasonics sonochemistry (2015-07-19)
M Mostafaei, B Ghobadian, M Barzegar, A Banakar
ZUSAMMENFASSUNG
This paper evaluates and optimizes the continuous production of biodiesel from waste cooking oil. In this research work, methanol and potassium hydroxide were used as catalyst engaging response surface methodology. For this purpose, the central composite experimental design (CCED), the effects of various factors such as irradiation distance, probe diameter, ultrasonic amplitude, vibration pulse and material flow into the reactor on reaction yield were studied to optimize the process. The results showed that all of the considered parameters affect the reaction efficiency significantly. The optimum combination of the findings include: irradiation distance which was 75 mm, probe diameter of 28 mm, ultrasonic amplitude of 56%, vibration pulse of 62% and flow rate of 50 ml/min that caused the reaction yield of 91.6% and energy consumption of 102.8 W. To verify this optimized combination, three tests were carried out. The results showed an average efficiency of 91.12% and 102.4 W power consumption which is well matched with the model's predictions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Schwefelsäure, 99.999%
Sigma-Aldrich
α-Linoleic acid, ≥98%
Sigma-Aldrich
Isopropylalkohol, ≥99.7%, FCC, FG
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Phenolphthalein, ACS reagent
Sigma-Aldrich
Linolsäure, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Heptan, anhydrous, 99%
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Phenolphthalein -Lösung, 0.5 wt. % in ethanol: water (1:1)
Sigma-Aldrich
Kaliumhydroxid, ≥99.95% trace metals basis
Sigma-Aldrich
Linolsäure, technical, 58-74% (GC)
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.