Merck
Moisture Barrier Composites Made of Non-Oxidized Graphene Flakes.
Small (Weinheim an der Bergstrasse, Germany) (2015-03-03)
Jungmo Kim, Sung Ho Song, Hyeon-Gyun Im, Gabin Yoon, Dongju Lee, Chanyong Choi, Jin Kim, Byeong-Soo Bae, Kisuk Kang, Seokwoo Jeon
ZUSAMMENFASSUNG
Graphene flakes (GFs) with minimized defects and oxidation ratios are incorporated into polyethylene (PE) to enhance the moisture barrier. GFs produced involving solvothermal intercalation show extremely low oxidation rates (3.17%), and are noncovalently functionalized in situ, inducing strong hydrophobicity. The fabricated composite possesses the best moisture barrier performance reported for a polymer-graphene composite.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Chinolin, reagent grade, 98%