Merck
  • Tumors Escape CD4+ T-cell-Mediated Immunosurveillance by Impairing the Ability of Infiltrating Macrophages to Indirectly Present Tumor Antigens.
Tumors Escape CD4+ T-cell-Mediated Immunosurveillance by Impairing the Ability of Infiltrating Macrophages to Indirectly Present Tumor Antigens.
Cancer research (2015-06-04)
Anders Aune Tveita, Fredrik Schjesvold, Ole Audun Haabeth, Marte Fauskanger, Bjarne Bogen
ZUSAMMENFASSUNG
Tumors cells can escape cytotoxic CD8+ T cells by preventing MHC I display of tumor antigens. It is unknown how tumors evade CD4+ T-cell responses, but because many tumor cells lack MHC II expression, novel mechanisms would be required. We have investigated this issue in a model in which MHC II(NEG) myeloma cells secrete a monoclonal Ig containing a V region L chain (VL) epitope recognized by CD4+ T cells. Infiltrating macrophages process and present the secreted tumor antigen to Th1 cells, resulting in induction of macrophage cytotoxicity and apparent rejection of the tumor. Despite long-term tumor protection in VL-specific T-cell receptor transgenic mice, we here describe that some myeloma cells persisted in a dormant state and, eventually, formed expanding tumors. Escape tumor cells maintained their secretion of complete (H+L) monoclonal Ig with unchanged sequence, while secretion of surplus free L chain was severely diminished. Although free L chains were efficiently processed and presented by tumor-infiltrating macrophages to CD4+ T cells, complete (H+L) monoclonal Ig was not. Forced overexpression of free L chain secretion reinstated tumor rejection. These results show that tumors can escape CD4+ T-cell-mediated rejection by impairing indirect presentation of tumor antigen by infiltrating macrophages. This occurs through a novel mechanism of immunoediting, in which modulation of the quaternary structure of the secreted tumor-specific antigen reduces its immunogenicity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Thymidin, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumsalicylat, ReagentPlus®, ≥99.5% (titration)
Sigma-Aldrich
Thymidin, ≥99%
Sigma-Aldrich
Natriumsalicylat, 98.0-102.0% anhydrous basis, meets USP testing specifications
Sigma-Aldrich
Natriumsalicylat, ≥99.5% (HPLC), puriss. p.a.
Sigma-Aldrich
Thymidin, ≥99.0% (HPLC)
Sigma-Aldrich
Natriumsalicylat, puriss. p.a., reag. Ph. Eur., 99.5-101.0% (calc. to the dried substance)