Merck
  • Comparative Transcriptome Analysis of Differentially Expressed Genes and Signaling Pathways between XY and YY Testis in Yellow Catfish.
Comparative Transcriptome Analysis of Differentially Expressed Genes and Signaling Pathways between XY and YY Testis in Yellow Catfish.
PloS one (2015-08-05)
Junjie Wu, Shuting Xiong, Jing Jing, Xin Chen, Weimin Wang, Jian-Fang Gui, Jie Mei
ZUSAMMENFASSUNG
YY super-males have rarely been detected in nature and only been artificially created in some fish species including tilapia and yellow catfish (Pelteobagrusfulvidraco), which provides a promising model for testis development and spermatogenesis. In our previous study, significant differences in morphology and miRNA expression were detected between XY and YY testis of yellow catfish. Here, solexa sequencing technology was further performed to compare mRNA expression between XY and YY testis. Compared with unigenes expressed in XY testis, 1146 and 1235 unigenes have significantly higher and lower expression in YY testis, respectively. 605 differentially expressed unigenes were annotated to 1604 GO terms with 319 and 286 genes having relative higher expression in XY and YY testis. KEGG analysis suggested different levels of PI3K-AKT and G protein-coupled receptor (GPCR) signaling pathways between XY and YY testis. Down-regulation of miR-141/429 in YY testis was speculated to promote testis development and maturation, and several factors in PI3K-AKT and GPCR signaling pathways were found as predicted targets of miR-141/429, several of which were confirmed by dual-luciferase reporter assays. Our study provides a comparative transcriptome analysis between XY and YY testis, and reveals interactions between miRNAs and their target genes that are possibly involved in regulating testis development and spermatogenesis.