Merck
Metal Domain Size Dependent Electrical Transport in Pt-CdSe Hybrid Nanoparticle Monolayers.
ACS nano (2015-06-09)
Michaela Meyns, Svenja Willing, Hauke Lehmann, Christian Klinke
ZUSAMMENFASSUNG
Thin films prepared of semiconductor nanoparticles are promising for low-cost electronic applications such as transistors and solar cells. One hurdle for their breakthrough is their notoriously low conductivity. To address this, we precisely decorate CdSe nanoparticles with platinum domains of one to three nanometers in diameter by a facile and robust seeded growth method. We demonstrate the transition from semiconductor to metal dominated conduction in monolayered films. By adjusting the platinum content in such solution-processable hybrid, oligomeric nanoparticles the dark currents through deposited arrays become tunable while maintaining electronic confinement and photoconductivity. Comprehensive electrical measurements allow determining the reigning charge transport mechanisms.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Selen, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Trioctylphosphin, 97%
Sigma-Aldrich
Selen, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Trioctylphosphin, technical grade, 90%
Sigma-Aldrich
Stickstoff, ≥99.998%
Sigma-Aldrich
Octadecylphosphonsäure, 97%
Sigma-Aldrich
Selen, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Selen, pellets, <5 mm, ≥99.99% trace metals basis
Selen, foil, 25x25mm, thickness 3mm, 99.95%
Selen, pellets, < 5mm, ≥99.999%