Merck
  • Magnetic graphene - polystyrene sulfonic acid nano composite: A dispersive cation exchange sorbent for the enrichment of aminoalcohols and ethanolamines from environmental aqueous samples.
Magnetic graphene - polystyrene sulfonic acid nano composite: A dispersive cation exchange sorbent for the enrichment of aminoalcohols and ethanolamines from environmental aqueous samples.
Journal of chromatography. A (2015-11-12)
Sridhar Chinthakindi, Ajay Purohit, Varoon Singh, Vijay Tak, D K Dubey, Deepak Pardasani
ZUSAMMENFASSUNG
Present study aimed at graphene surface modification to achieve selective analyte binding in dispersive solid phase extraction. Magnetic graphene - polystyrene sulfonic acid (MG-PSS) cation exchange nano-composite was prepared by non-covalent wrapping method. Composite was characterized by FT-IR and zeta potential. Material exhibited good dispersion in water and high exchange capacity of 1.97±0.16mMg(-1). Prepared nano-sorbent was then exploited for the cation exchange extraction and gas chromatography mass spectrometric analysis of Chemical Weapons Convention relevant aminoalcohols and ethanolamines from aqueous samples. Extraction parameters such as sorbent amount, extraction time, desorption conditions and sample pH were optimized and effect of common matrix interferences such as polyethylene glycol and metal salts was also studied. Three milligram of sorbent per mL of sample with 20min of extraction time at room temperature afforded 70-81% recoveries of the selected analytes spiked at concentration level of 1μgmL(-1). Method showed good linearity in the studied range with r(2)≥0.993. The limits of detection and limits of quantification ranged from 23 to 54ngmL(-1) and 72 to 147ngmL(-1), respectively. The relative standard deviation for intra- and inter-day precision ranged from 4.6 to 10.2% and 7.4 to 14.8% respectively. Applicability of the method to different environmental samples as well as the proficiency tests conducted by the Organization for the Prohibition of Chemical Weapons (OPCW) was also ascertained.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Kaliumpermanganat, ACS reagent, ≥99.0%
Sigma-Aldrich
Triethylamin, ≥99.5%
Sigma-Aldrich
Kaliumpersulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Triethanolamin, ≥99.0% (GC)
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Triethylamin, ≥99%
Supelco
N,O-Bis-(trimethylsilyl)-trifluoracetamid mit Trimethylchlorsilan, with 1% trimethylchlorosilane, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Triethylamin, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamin, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
N,O-Bis(trimethylsilyl)trifluoracetamid, ≥99%
Sigma-Aldrich
Kaliumpermanganat, ACS reagent, ≥99.0%, low in mercury
Sigma-Aldrich
Hydrazin Monohydrat, N2H4 64-65 %, reagent grade, ≥97%
Sigma-Aldrich
Hydrazin Hydrat, reagent grade, N2H4 50-60 %
Sigma-Aldrich
Triethanolamin, reagent grade, 98%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrazinhydrat -Lösung, puriss. p.a., 24-26% in H2O (RT)
Sigma-Aldrich
Triethoxysilan, 95%
Sigma-Aldrich
Kaliumpersulfat, 99.99% trace metals basis
Sigma-Aldrich
Kaliumpermanganat, 97%
Sigma-Aldrich
2-(Diethylamino)ethanol, ≥99.5%
Sigma-Aldrich
N,N-Diethylanilin, ≥99%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 34.5-36.5%
Sigma-Aldrich
Triethylamin, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
3-Chinuclidinol, 99%
Sigma-Aldrich
Triethanolamin, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamin, ≥99.5%
Sigma-Aldrich
Triethanolamin, puriss. p.a., ≥99% (GC)
Sigma-Aldrich
Triethylamin, ampule, ≥99.5% (GC)