Merck
  • Selective enrichment and determination of monoamine neurotransmitters by CU(II) immobilized magnetic solid phase extraction coupled with high-performance liquid chromatography-fluorescence detection.
Selective enrichment and determination of monoamine neurotransmitters by CU(II) immobilized magnetic solid phase extraction coupled with high-performance liquid chromatography-fluorescence detection.
Talanta (2015-11-26)
Maofang He, Chaozhan Wang, Yinmao Wei
ZUSAMMENFASSUNG
In this paper, iminodiacetic acid-Cu(II) functionalized Fe3O4@SiO2 magnetic nanoparticles were prepared and used as new adsorbents for magnetic solid phase extraction (MSPE) of six monoamine neurotransmitters (MNTs) from rabbit plasma. The selective enrichment of MNTs at pH 5.0 was motivated by the specific coordination interaction between amino groups of MNTs and the immobilized Cu(II). The employed weak acidic extraction condition avoided the oxidation of MNTs, and thus facilitated operation and ensured higher recoveries. Under optimal conditions, the recoveries of six MNTs from rabbit plasma were in the range of 83.9-109.4%, with RSD of 2.0-10.0%. When coupled the Cu(II) immobilized MSPE with high-performance liquid chromatography-fluorescence detection, the method exhibited relatively lower detection limits than the previously reported methods, and the method was successfully used to determine the endogenous MNTs in rabbit plasma. The proposed method has potential application for the determination of MNTs in biological samples. Also, the utilization of coordination interaction to improve the selectivity might open another way to selectively enrich small alkaloids from complex samples.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
Tetraethylorthosilikat, reagent grade, 98%
Sigma-Aldrich
Dopamin -hydrochlorid
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Salzsäure, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Salzsäure, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Tetraethylorthosilikat, ≥99.0% (GC)
Sigma-Aldrich
Chlorwasserstoff -Lösung, 2.0 M in diethyl ether
Sigma-Aldrich
Serotonin -hydrochlorid, powder
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Chlorwasserstoff -Lösung, 1.0 M in diethyl ether
Sigma-Aldrich
Salzsäure -Lösung, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Tetraethylorthosilikat, 99.999% trace metals basis
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
DL-Isoproterenol -hydrochlorid
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Salzsäure -Lösung, 32 wt. % in H2O, FCC
Sigma-Aldrich
Chlorwasserstoff -Lösung, 1.0 M in acetic acid
Sigma-Aldrich
Tetraethylorthosilicat, packaged for use in deposition systems