Merck
  • Prolonged glutamate excitotoxicity increases GluR1 immunoreactivity but decreases mRNA of GluR1 and associated regulatory proteins in dissociated rat retinae in vitro.
Prolonged glutamate excitotoxicity increases GluR1 immunoreactivity but decreases mRNA of GluR1 and associated regulatory proteins in dissociated rat retinae in vitro.
Biochimie (2015-03-21)
Michael Challenor, Ryan O'Hare Doig, Paula Fuller, Marcus Giacci, Carole Bartlett, Celeste H Wale, Greg S Cozens, Livia Hool, Sarah Dunlop, K Swaminathan Iyer, Jennifer Rodger, Melinda Fitzgerald
ZUSAMMENFASSUNG
Glutamate excitotoxicity contributes to damage following injury to the central nervous system via mechanisms including changes in the expression of receptors, calcium overload, oxidative stress and cell death. Excitotoxicity is triggered by glutamate binding to receptors, including calcium permeable AMPA receptors, which can be upregulated under injury conditions. However, the transcriptional response of AMPA receptor regulatory proteins to excitotoxic conditions is unknown. Here, we use real-time quantitative PCR (RT-qPCR), to determine the effect of prolonged glutamate excitotoxicity on the expression of mRNA encoding for GluR1 and AMPA receptor regulatory proteins in dissociated rat retinal cultures that include neuronal (retinal ganglion cell (RGCs)) and glial (Müller) cell populations. mRNA levels of GluR1 and regulators of the GluR1 subunit of AMPA receptors, including Sap97, Cnih2 and Cnih3, decreased following 6, 24 and 48 h incubation with 5 mM glutamate: related regulators not associated with GluR1 were unaffected. In contrast, GluR1 protein, assessed immunohistochemically, was increased in both RGCs and Müller cells after 24 h glutamate exposure: western blotting analysis was inconclusive. Reductions in mRNA of GluR1 and associated regulators occurred prior to cell death, which was first detected at 24 h, and substantial by 48 h. Exposure to glutamate acutely increased the intracellular calcium concentration in the cultures and by 24 h, reactive oxygen species were increased. Our data suggest a negative feedback mechanism in retinal cells, that down-regulates transcription of genes encoding GluR1 regulatory proteins in response to glutamate exposure. Despite this mechanism, GluR1 protein levels remain increased, and are associated with increased reactive species and cell death. Therapeutic strategies targeting calcium permeable AMPA receptors should take into account that increases in GluR1 protein are not necessarily associated with increases in associated mRNA levels over time.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
PIPES, ≥99% (titration)
Sigma-Aldrich
Saccharose, ACS reagent
SAFC
L-Glutamin
Sigma-Aldrich
Saccharose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
PIPES, BioPerformance Certified, suitable for cell culture
Sigma-Aldrich
Calcein-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
Sigma-Aldrich
Mangan, powder, ≥99.9% trace metals basis
Sigma-Aldrich
Saccharose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 34.5-36.5%
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Ethidiumhomodimer, suitable for fluorescence, ~90% (HPCE)
Sigma-Aldrich
Saccharose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Mangan, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Saccharose, meets USP testing specifications
Sigma-Aldrich
PIPES, BioXtra, ≥99% (titration)
Sigma-Aldrich
PIPES, BioXtra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥90.0% (HPLC)
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Sigma-Aldrich
DL-Cystein, technical grade
Sigma-Aldrich
2,2′-Bichinolin-4,4′-dicarbonsäure Dinatriumsalz Dihydrat, ≥98% (HPLC)