Merck
  • Influence of the amount of co-infused amino acids on post-therapeutic potassium levels in peptide receptor radionuclide therapy.
Influence of the amount of co-infused amino acids on post-therapeutic potassium levels in peptide receptor radionuclide therapy.
EJNMMI research (2014-01-01)
Constantin Lapa, Rudolf A Werner, Christina Bluemel, Katharina Lückerath, Andreas Schirbel, Alexander Strate, Andreas K Buck, Ken Herrmann
ZUSAMMENFASSUNG
Peptide receptor radionuclide therapy (PRRT) is routinely used for advanced or metastasized neuroendocrine tumours (NET). To prevent nephrotoxicity, positively charged amino acids (AA) are co-infused. The aim of this study was to correlate the risk for therapy-related hyperkalaemia with the total amount of AA infused. Twenty-two patients undergoing PRRT with standard activities of (177)Lu-DOTATATE/-TOC were monitored during two following treatment cycles with co-infusion of 75 and 50 g of AA (l-arginine and l-lysine), respectively. Mean serum levels of potassium and other parameters (glomerular filtration rate [GFR], creatinine, blood urea nitrogen [BUN], phosphate, chloride, lactate dehydrogenase) prior to, 4 h and 24 h after AA infusion were compared. Self-limiting hyperkalaemia (>5.0 mmol/l) resolving after 24 h occurred in 91% (20/22) of patients in both protocols. Potassium levels, BUN, creatinine, GFR, phosphate, chloride and LDH showed a similar range at 4 h after co-infusion of 75 or 50 g of AA, respectively (p?>?0.05). Only GFR and creatinine levels at 24 h varied significantly between the two co-infusion protocols (p?<?0.05). Hyperkalaemia is a frequent side effect of AA infusion in PRRT. Varying the dose of co-infused amino acids did not impact on the incidence and severity of hyperkalaemia.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Salzsäure -Lösung, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Kalium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Kaliumhydrid, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Salzsäure -Lösung, 32 wt. % in H2O, FCC
Sigma-Aldrich
Octreotide, ≥98% (HPLC)
Sigma-Aldrich
Kaliumhydrid, in paraffin