Merck
  • Hydrogen Peroxide Induce Human Cytomegalovirus Replication through the Activation of p38-MAPK Signaling Pathway.
Hydrogen Peroxide Induce Human Cytomegalovirus Replication through the Activation of p38-MAPK Signaling Pathway.
Viruses (2015-06-09)
Jun Xiao, Jiang Deng, Liping Lv, Qiong Kang, Ping Ma, Fan Yan, Xin Song, Bo Gao, Yanyu Zhang, Jinbo Xu
ZUSAMMENFASSUNG
Human cytomegalovirus (HCMV) is a major risk factor in transplantation and AIDS patients, which induces high morbidity and mortality. These patients infected with HCMV experience an imbalance of redox homeostasis that cause accumulation of reactive oxygen species (ROS) at the cellular level. H2O2, the most common reactive oxygen species, is the main byproduct of oxidative metabolism. However, the function of H2O2 on HCMV infection is not yet fully understood and the effect and mechanism of N-acetylcysteine (NAC) on H2O2-stimulated HCMV replication is unclear. We, therefore, examined the effect of NAC on H2O2-induced HCMV production in human foreskin fibroblast cells. In the present study, we found that H2O2 enhanced HCMV lytic replication through promoting major immediate early (MIE) promoter activity and immediate early (IE) gene transcription. Conversely, NAC inhibited H2O2-upregulated viral IE gene expression and viral replication. The suppressive effect of NAC on CMV in an acute CMV-infected mouse model also showed a relationship between antioxidants and viral lytic replication. Intriguingly, the enhancement of HCMV replication via supplementation with H2O2 was accompanied with the activation of the p38 mitogen-activated protein kinase pathway. Similar to NAC, the p38 inhibitor SB203580 inhibited H2O2-induced p38 phosphorylation and HCMV upregulation, while upregulation of inducible ROS was unaffected. These results directly relate HCMV replication to H2O2, suggesting that treatment with antioxidants may be an attractive preventive and therapeutic strategy for HCMV.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Acetyl-L-Cystein, BioReagent, suitable for cell culture
Sigma-Aldrich
N-Acetyl-L-Cystein, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
Luminol, 97%
Sigma-Aldrich
3-Amino-1,2,4-triazol, ≥95% (TLC)
Sigma-Aldrich
Luminol, ≥97% (HPLC)
Sigma-Aldrich
N-Acetyl-L-Cystein, BioXtra, ≥99% (TLC)
Sigma-Aldrich
Aurintri-Carboxylsäure, practical grade, ≥85% (titration), powder
Sigma-Aldrich
Aurintri-Carboxylsäure Ammoniumsalz, ACS reagent
Sigma-Aldrich
Aurintri-Carboxylsäure Ammoniumsalz, powder
Sigma-Aldrich
4H-1,2,4-Triazol-3-amine, AldrichCPR
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)