Merck
Faster through training: The anammox case.
Water research (2015-06-16)
T Lotti, R Kleerebezem, J M Abelleira-Pereira, B Abbas, M C M van Loosdrecht
ZUSAMMENFASSUNG
Anaerobic ammonium oxidizing (anammox) bacteria based technologies are widely applied for nitrogen removal from warm (25-40 °C) wastewater with high ammonium concentrations (∼1 gNH4-N L(-1)). Extension of the operational window of this energy and resource efficient process is restricted by the "supposed" low growth rate of the responsible microorganisms. Here we demonstrate that the maximum specific growth rate (μ(max)) of anammox bacteria can be increased to a μ(max) value of 0.33 d(-1) by applying a novel selection strategy based on the maximization of the electron transfer capacity in a membrane bioreactor. This value is four times higher than the highest previously reported value. The microbial community was strongly dominated by anammox bacteria closely related (99%) to Candidatus Brocadia sp.40 throughout the experiment. The results described here demonstrate the remarkable capacity of a phylogenetically stable anammox community to adjust its growth rate in response to a change in the cultivation conditions imposed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Harnstoff, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Harnstoff, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Formamid, ≥99.5% (GC), BioReagent, Molecular Biology
Supelco
Harnstoff, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Formamid, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Harnstoff -Lösung, BioUltra, ~8 M in H2O
Sigma-Aldrich
Harnstoff, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Harnstoff, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Harnstoff, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Sigma-Aldrich
Harnstoff, suitable for electrophoresis
Sigma-Aldrich
Harnstoff, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Essigsäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Essigsäure, natural, ≥99.5%, FG
Sigma-Aldrich
Harnstoff, meets USP testing specifications
Sigma-Aldrich
Harnstoff, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Harnstoff -Lösung, 40 % (w/v) in H2O
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, purum, ≥99.0% (TLC)
Sigma-Aldrich
Essigsäure-12C2, 99.9 atom % 12C
Sigma-Aldrich
Harnstoff-12C, 99.9 atom % 12C
Sigma-Aldrich
Formamid -Lösung, suitable for NMR (reference standard), 90% in DMSO-d6 (99.9 atom % D), NMR tube size 10 mm × 8 in.
Supelco
Formamid -Lösung, suitable for NMR (reference standard), 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.