Merck
  • The utility of cardiac biomarkers and echocardiography for the early detection of bevacizumab- and sunitinib-mediated cardiotoxicity.
The utility of cardiac biomarkers and echocardiography for the early detection of bevacizumab- and sunitinib-mediated cardiotoxicity.
American journal of physiology. Heart and circulatory physiology (2015-06-21)
Kimberly-Ann Bordun, Sheena Premecz, Megan daSilva, Soma Mandal, Vineet Goyal, Tamara Glavinovic, Matthew Cheung, David Cheung, Christopher W White, Rakesh Chaudhary, Darren H Freed, Hector R Villarraga, Joerg Herrmann, Manish Kohli, Amir Ravandi, James Thliveris, Marshall Pitz, Pawan K Singal, Sharon Mulvagh, Davinder S Jassal
ZUSAMMENFASSUNG
The recent introduction of novel anticancer therapies, including bevacizumab (BVZ) and sunitinib (SNT), is associated with an increased risk of cardiotoxicity. However, early identification of left ventricular (LV) systolic dysfunction may facilitate dose modification and avoid the development of advanced heart failure. Using a murine model of BVZ- and SNT-mediated cardiotoxicity, we investigated whether cardiac biomarkers and/or tissue velocity imaging (TVI) using echocardiography can detect early changes in cardiac function, before a decrease in LV ejection fraction is identified. A total of 75 wild-type C57Bl/6 male mice were treated with either 0.9% saline, BVZ, or SNT. Serial monitoring of blood pressure, high-sensitivity troponin I, and echocardiographic indexes were performed over a 14-day study period, after which the mice were euthanized for histological and biochemical analyses. Mice treated with either BVZ or SNT developed systemic hypertension as early as day 7, which increased by day 14. Cardiac biomarkers, specifically high-sensitivity troponin I, were not predictive of early LV systolic dysfunction. Although conventional LV ejection fraction values decreased at day 13 in mice treated with either BVZ or SNT, TVI confirmed early LV systolic dysfunction at day 8. Histological and biochemical analysis demonstrated loss of cellular integrity, increased oxidative stress, and increased cardiac apoptosis in mice treated with BVZ or SNT therapy at day 14. In a murine model of BVZ- or SNT-mediated cardiomyopathy, noninvasive assessment by TVI detected early LV systolic dysfunction before alterations in conventional echocardiographic indexes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Stickstoff, ≥99.998%
Sigma-Aldrich
Natriumdodecylsulfat, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Natriumdodecylsulfat, ≥90% ((Assay))
Sigma-Aldrich
Natriumdodecylsulfat, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri