Merck
  • Simultaneous determination of cobalt and nickel in vitamin B12 samples using high-resolution continuum source atomic absorption spectrometry.
Simultaneous determination of cobalt and nickel in vitamin B12 samples using high-resolution continuum source atomic absorption spectrometry.
Talanta (2015-11-26)
Franciele Rovasi Adolfo, Paulo Cícero do Nascimento, Denise Bohrer, Leandro Machado de Carvalho, Carine Viana, Ananda Guarda, Alexsandro Nunes Colim, Patricia Mattiazzi
ZUSAMMENFASSUNG
Nickel and cobalt were simultaneously assayed in vitamin B12 formulations by using atomic spectrometry. The proposed method is based on a compromise between the proximity of specific Ni and Co spectral lines and the relative abundances of the analytes in the samples. The analytes were found in concentrations ranging from 9.48 to 26.20µg L(-1) (Ni) and from 156.90 to 279.25mg L(-1) (Co) in the commercial samples of vitamin B12. The limits of detection and quantification were 1.21 and 3.64mg L(-1) for Co and 0.39 and 1.19µg L(-1) for Ni. Sample cleanup was not necessary for the determinations, and the interferences were discussed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salpetersäure, ACS reagent, 70%
Sigma-Aldrich
Salpetersäure, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Salpetersäure, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Salpetersäure, red, fuming, HNO3 >90 %
Sigma-Aldrich
Salpetersäure, ACS reagent, ≥90.0%
Sigma-Aldrich
Salpeter-14N-Säurelösung, ~10 N in H2O, 99.99 atom % 14N