Merck
Rapid in vivo apparent diffusion coefficient mapping of hyperpolarized (13) C metabolites.
Magnetic resonance in medicine (2014-09-13)
Bertram L Koelsch, Galen D Reed, Kayvan R Keshari, Myriam M Chaumeil, Robert Bok, Sabrina M Ronen, Daniel B Vigneron, John Kurhanewicz, Peder E Z Larson
ZUSAMMENFASSUNG
Hyperpolarized (13) C magnetic resonance allows for the study of real-time metabolism in vivo, including significant hyperpolarized (13) C lactate production in many tumors. Other studies have shown that aggressive and highly metastatic tumors rapidly transport lactate out of cells. Thus, the ability to not only measure the production of hyperpolarized (13) C lactate but also understand its compartmentalization using diffusion-weighted MR will provide unique information for improved tumor characterization. We used a bipolar, pulsed-gradient, double spin echo imaging sequence to rapidly generate diffusion-weighted images of hyperpolarized (13) C metabolites. Our methodology included a simultaneously acquired B1 map to improve apparent diffusion coefficient (ADC) accuracy and a diffusion-compensated variable flip angle scheme to improve ADC precision. We validated this sequence and methodology in hyperpolarized (13) C phantoms. Next, we generated ADC maps of several hyperpolarized (13) C metabolites in a normal rat, rat brain tumor, and prostate cancer mouse model using both preclinical and clinical trial-ready hardware. ADC maps of hyperpolarized (13) C metabolites provide information about the localization of these molecules in the tissue microenvironment. The methodology presented here allows for further studies to investigate ADC changes due to disease state that may provide unique information about cancer aggressiveness and metastatic potential.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Brenztraubensäure, 98%
Sigma-Aldrich
Brenztraubensäure, ≥97%, FG
Sigma-Aldrich
Brenztraubensäure, 95%
Sigma-Aldrich
Brenztraubensäure, natural, ≥95%, FG