Merck
  • Chiral Inhibition of Rivaroxaban Derivatives Towards UDP-Glucuronosyltransferase (UGT) Isoforms.
Chiral Inhibition of Rivaroxaban Derivatives Towards UDP-Glucuronosyltransferase (UGT) Isoforms.
Chirality (2015-10-02)
Zhuhua Yao, Yong-Zhe Liu, Ai-Lun Ma, Shu-Fen Wang, Dan Lu, Cui-Min Hu, Yan-Yan Zhang, Haina Wang, Lingyun Hu, Jun Deng, Kun Yang, Zhong-Ze Fang
ZUSAMMENFASSUNG
Rivaroxaban is an oral direct factor Xa (FXa) inhibitor clinically used to prevent and treat thromboembolic disorders. Drug-drug interaction (DDI) exist for rivaroxaban and the inhibitors of CYP3A4/5. This study aims to investigate the inhibition of rivaroxaban and its derivatives with a chiral center towards UDP-glucuronosyltransferases (UGTs). Chemical synthesis was performed to obtain rivaroxaban derivatives with different chiral centers. UGTs supersomes-catalyzed 4-methylumbelliferone (4-MU) glucuronidation was employed to evaluate the inhibition potential towards various UGT isoforms. A significant influence of rivaroxaban derivatives towards UGT1A3 was observed. Chiral centers produce different effects towards the effect of four pairs of rivaroxaban derivatives towards UGT1A3 activity, with stronger inhibition potential of S1 than R1, but stronger inhibition capability of R2, R3, R4 than S2, S3, and S4. Competitive inhibition of R3 and R4 towards UGT1A3 was demonstrated by Dixon and Lineweaver-Burk plots. In conclusion, the significant influence of rivaroxaban derivatives towards UGT1A3's activity was demonstrated in the present study. The chirality centers highly affected the inhibition behavior of rivaroxaban derivatives towards UGT1A3.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Tetramethylsilan, NMR grade, ACS reagent, ≥99.9%
Sigma-Aldrich
4-Methyl-umbelliferon, ≥98%
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Umbelliferon, 99%
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Uridin-5′-diphosphoglucuronsäure Ammoniumsalz, 98-100%
Sigma-Aldrich
Tetramethylsilan, ≥99.0% (GC)
Sigma-Aldrich
4-Methylumbelliferyl β-D-Galactopyranosid, ≥99% (TLC)
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Umbelliferon, suitable for fluorescence indicator, ≥98.0% (HPLC)
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Tetramethylsilan, electronic grade, ≥99.99% trace metals basis
Sigma-Aldrich
Chloroform-d, 99.8 atom % D