Merck
  • Evidence of a Native Northwest Atlantic COI Haplotype Clade in the Cryptogenic Colonial Ascidian Botryllus schlosseri.
Evidence of a Native Northwest Atlantic COI Haplotype Clade in the Cryptogenic Colonial Ascidian Botryllus schlosseri.
The Biological bulletin (2015-07-01)
Philip O Yund, Catherine Collins, Sheri L Johnson
ZUSAMMENFASSUNG
The colonial ascidian Botryllus schlosseri should be considered cryptogenic (i.e., not definitively classified as either native or introduced) in the Northwest Atlantic. Although all the evidence is quite circumstantial, over the last 15 years most research groups have accepted the scenario of human-mediated dispersal and classified B. schlosseri as introduced; others have continued to consider it native or cryptogenic. We address the invasion status of this species by adding 174 sequences to the growing worldwide database for the mitochondrial gene cytochrome c oxidase subunit I (COI) and analyzing 1077 sequences to compare genetic diversity of one clade of haplotypes in the Northwest Atlantic with two hypothesized source regions (the Northeast Atlantic and Mediterranean). Our results lead us to reject the prevailing view of the directionality of transport across the Atlantic. We argue that the genetic diversity patterns at COI are far more consistent with the existence of at least one haplotype clade in the Northwest Atlantic (and possibly a second) that substantially pre-dates human colonization from Europe, with this native North American clade subsequently introduced to three sites in Northeast Atlantic and Mediterranean waters. However, we agree with past researchers that some sites in the Northwest Atlantic have more recently been invaded by alien haplotypes, so that some populations are currently composed of a mixture of native and invader haplotypes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%