Merck
  • Lactobacillus crispatus inhibits growth of Gardnerella vaginalis and Neisseria gonorrhoeae on a porcine vaginal mucosa model.
Lactobacillus crispatus inhibits growth of Gardnerella vaginalis and Neisseria gonorrhoeae on a porcine vaginal mucosa model.
BMC microbiology (2015-12-15)
Laura M Breshears, Vonetta L Edwards, Jacques Ravel, Marnie L Peterson
ZUSAMMENFASSUNG
The vaginal microbiota can impact the susceptibility of women to bacterial vaginosis (BV) and sexually transmitted infections (STIs). BV is characterized by depletion of Lactobacillus spp., an overgrowth of anaerobes (often dominated by Gardnerella vaginalis) and a pH > 4.5. BV is associated with an increased risk of acquiring STIs such as chlamydia and gonorrhea. While these associations have been identified, the molecular mechanism(s) driving the risk of infections are unknown. An ex vivo porcine vaginal mucosal model (PVM) was developed to explore the mechanistic role of Lactobacillus spp. in affecting colonization by G. vaginalis and Neisseria gonorrhoeae. The data presented here demonstrate that all organisms tested can colonize and grow on PVM to clinically relevant densities. Additionally, G. vaginalis and N. gonorrhoeae form biofilms on PVM. It was observed that lactic acid, acetic acid, and hydrochloric acid inhibit the growth of G. vaginalis on PVM in a pH-dependent manner. N. gonorrhoeae grows best in the presence of lactic acid at pH 5.5, but did not grow well at this pH in the presence of acetic acid. Finally, a clinical Lactobacillus crispatus isolate (24-9-7) produces lactic acid and inhibits growth of both G. vaginalis and N. gonorrhoeae on PVM. These data reveal differences in the effects of pH, various acids and L. crispatus on the growth of G. vaginalis and N. gonorrhoeae on a live vaginal mucosal surface. The PVM is a useful model for studying the interactions of commensal vaginal microbes with pathogens and the mechanisms of biofilm formation on the vaginal mucosa.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumphosphat, ACS reagent, ≥99.0%
Sigma-Aldrich
Iod, ACS reagent, ≥99.8%, solid
Sigma-Aldrich
Natriumthiosulfat, ReagentPlus®, 99%
Sigma-Aldrich
Kaliumphosphat, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Natriumthiosulfat, purum p.a., anhydrous, ≥98.0% (RT)
Sigma-Aldrich
Iod, flakes, ReagentPlus®, ≥99%
Sigma-Aldrich
Milchsäure, meets USP testing specifications
Supelco
Iod, ReagentPlus®, ≥99.8% (titration)
Sigma-Aldrich
Kaliumphosphat, ReagentPlus®
Sigma-Aldrich
Milchsäure, 88%, FCC
Sigma-Aldrich
Kaliumphosphat, Molecular Biology, ≥98.0%
Sigma-Aldrich
Milchsäure, natural, ≥85%
Sigma-Aldrich
Milchsäure -Lösung, ACS reagent, ≥85%
Sigma-Aldrich
Iod, ≥99.99% trace metals basis
Sigma-Aldrich
Natriumthiosulfat, ≥99.99% trace metals basis
Sigma-Aldrich
Iod, 99.999% trace metals basis
Sigma-Aldrich
Tantal(V)-ethoxid, 99.98% trace metals basis
Sigma-Aldrich
Kaliumphosphat, BioUltra, Molecular Biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Azodicarbonsäure-diethylester -Lösung, purum, ~40% in toluene (H-NMR)
Sigma-Aldrich
DL-Milchsäure, 85 % (w/w), syrup
Sigma-Aldrich
Iod, anhydrous, beads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Iod, ReagentPlus®, 99.7% trace metals basis, beads, 1-3 mm
Sigma-Aldrich
Kaliumphosphat, 99.99% trace metals basis
Sigma-Aldrich
Natriumthiosulfat -Lösung, 2 g/dL in deionized water
Tantal(V)-ethoxid, packaged for use in deposition systems