Merck
  • Induction of a Th1 immune response and suppression of IgE via immunotherapy with a recombinant hybrid molecule encapsulated in liposome-protamine-DNA nanoparticles in a model of experimental allergy.
Induction of a Th1 immune response and suppression of IgE via immunotherapy with a recombinant hybrid molecule encapsulated in liposome-protamine-DNA nanoparticles in a model of experimental allergy.
Immunologic research (2015-05-11)
Hamid Reza Nouri, Abdolreza Varasteh, Mahmoud Reza Jaafari, Janet M Davies, Mojtaba Sankian
ZUSAMMENFASSUNG
Liposome-protamine-DNA nanoparticles (LPD) are safe, effective, and non-toxic adjuvants that induce Th1-like immune responses. We hypothesized that encapsulation of allergens into liposomes could be an appropriate option for immunotherapy. The present study evaluated the immunotherapeutic potential of a recombinant hybrid molecule (rHM) encapsulated in LPD nanoparticles in a murine model of Chenopodium album allergy. BALB/c mice were sensitized with the allergen in alum, and the immunotherapy procedure was performed by subcutaneous injections of LPD-rHM, rHM, or empty LPD at weekly intervals. Sensitized mice developed a Th2-biased immune response characterized by strong specific IgG1 and IgE production, IL-4, and the transcription factor GATA3 in spleen cell cultures. Treatment with the LPD-rHM resulted in a reduction in IgE and a marked increase in IgG2a. The LPD-rHM induced allergen-specific responses with relatively high interferon-gamma production, as well as expression of the transcription factor T-bet in stimulated splenocytes. In addition, lymphoproliferative responses were higher in the LPD-rHM-treated mice than in the other groups. Removal of the nanoparticles from the rHM resulted in a decrease in the allergen's immunogenicity. These results indicate that the rHM complexed with LPD nanoparticles has a marked suppressive effect on the allergic response and caused a shift toward a Th1 pathway.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumbicarbonat, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Natriumcarbonat, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Natriumcarbonat, BioXtra, ≥99.0%
Sigma-Aldrich
Natriumbicarbonat, BioXtra, 99.5-100.5%
Sigma-Aldrich
Natriumcarbonat, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Aluminium-Kaliumsulfat Dodecahydrat, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Natriumbicarbonat-12C, 13C-depleted, 99.9 atom % 12C
Sigma-Aldrich
Aluminium-Kaliumsulfat Dodecahydrat, BioXtra, ≥98.0%
Sigma-Aldrich
Natriumcarbonat-12C, 99.9 atom % 12C