Merck
  • Efficacy of Change to New P2Y12 Receptor Antagonists in Patients High on Treatment Platelet Reactivity Undergoing Percutaneous Coronary Intervention.
Efficacy of Change to New P2Y12 Receptor Antagonists in Patients High on Treatment Platelet Reactivity Undergoing Percutaneous Coronary Intervention.
Clinical and applied thrombosis/hemostasis : official journal of the International Academy of Clinical and Applied Thrombosis/Hemostasis (2014-12-20)
Rogelio Robledo-Nolasco, A Godínez-Montes de Oca, J F Zaballa-Contreras, J A Suárez-Cuenca, P Mondragón-Terán, A F Rubio-Guerra, M A Meléndez-Alcántara
ZUSAMMENFASSUNG
Selective intensification of platelet inhibition may improve high on treatment platelet reactivity (HPR). We evaluated the efficacy of dual-antiplatelet therapy, including clopidogrel (CPG), compared to new P2Y12-receptor antagonists in patients with HPR undergoing percutaneous coronary intervention, regarding the outcome of composite major adverse cardiac events (MACEs, including death, acute coronary syndrome [ACS], and stent restenosis). The presence of HPR (71 of 181 patients) almost doubled the risk of MACEs. The new antiplatelet agent reduced MACEs (45.8%, 26%, and 16.7% for CPG, prasugrel, and ticagrelor [TGL]; RR 0.36; 0.13-0.98, P = .03, TGL), specifically in patients with ACS. Failure to reduce HPR after the antiplatelet change and diabetes were independent predictors for MACEs. The HPR was early and effectively reduced after changing the antiplatelet therapy, but the intensity of this reduction did not significantly decrease the risk of MACEs. These findings support the benefit of HPR-guided intensification of platelet inhibition. Whether the intensity of this reduction improves the patient's clinical outcomes deserves further investigation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Zitrat-konzentrierte Lösung -Lösung, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Zitrat-konzentrierte Lösung -Lösung, BioUltra, Molecular Biology, 1 M in H2O