Merck
  • Direct degradation of cellulosic biomass to bio-hydrogen from a newly isolated strain Clostridium sartagoforme FZ11.
Direct degradation of cellulosic biomass to bio-hydrogen from a newly isolated strain Clostridium sartagoforme FZ11.
Bioresource technology (2015-05-27)
Jing-Nan Zhang, Yan-Hong Li, Hui-Qin Zheng, Yao-Ting Fan, Hong-Wei Hou
ZUSAMMENFASSUNG
A mesophilic hydrogen-producing strain, Clostridium sartagoforme FZ11, had been newly isolated from cow dung compost acclimated using microcrystalline cellulose (MCC) for at least 30 rounds in an anaerobic bioreactor, and identified by the 16S rDNA gene sequencing, which could directly utilized various carbon sources, especially cellulosic biomass, to produce hydrogen. The maximum hydrogen yields from MCC (10 g/l) and carboxymethyl cellulose (CMC, 10 g/l) were 77.2 and 64.6 ml/g, separately. Furthermore, some key parameters of affecting hydrogen production from raw corn stalk were also optimized. The maximal hydrogen yield and substrate degradation rate from raw corn stalk were 87.2 ml/g and 41.2% under the optimized conditions with substrate concentration of 15 g/l, phosphate buffer of 0.15 M, urea of 6 g/l and initial pH of 6.47 at 35 °C. The result showed that the strain FZ11 would be an ideal candidate to directly convert cellulosic biomass into bio-hydrogen without substrate pretreatment.