Merck
  • Antiviral effects of grape seed extract against feline calicivirus, murine norovirus, and hepatitis A virus in model food systems and under gastric conditions.
Antiviral effects of grape seed extract against feline calicivirus, murine norovirus, and hepatitis A virus in model food systems and under gastric conditions.
Food microbiology (2015-09-05)
Snehal S Joshi, Xiaowei Su, Doris H D'Souza
ZUSAMMENFASSUNG
Grape seed extract (GSE) has antiviral activities against hepatitis A virus (HAV) and human norovirus surrogates (feline calicivirus (FCV-F9) and murine norovirus (MNV-1)). The objectives of this study were to determine (1) time and dose-dependence of GSE against FCV-F9, MNV-1, and HAV at room temperature (RT) and 37 °C over 24 h; and (2) GSE effects in model foods (apple juice (AJ) and 2% milk) and simulated gastric conditions at 37 °C. Viruses at ∼5 log PFU/ml were treated with 0.5-8 mg/ml GSE prepared in water, AJ, milk or gastric juices, or water over 24 h at RT or 37 °C. Infectivity of triplicate treatments was evaluated using plaque assays. GSE effects increased with time and concentration. GSE at 1 mg/ml in AJ reduced MNV-1 to undetectable levels after 1 h and by 1 log in milk after 24 h. GSE at 1 and 2 mg/ml in AJ reduced HAV to undetectable levels after 1 h, while 2 and 4 mg/ml GSE in milk caused ∼1 log reduction after 24 h. GSE at 2 mg/ml in intestinal fluid reduced FCV-F9, MNV-1 and HAV to undetectable levels after 6 h. GSE appears to be a suitable natural option for foodborne viral reduction.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DL-Apfelsäure, ReagentPlus®, ≥99%
Sigma-Aldrich
Natriumcarbonat, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Natriumcarbonat, BioXtra, ≥99.0%
Sigma-Aldrich
DL-Apfelsäure, 99%
Sigma-Aldrich
DL-Apfelsäure, ≥98% (capillary GC)
Sigma-Aldrich
Natriumcarbonat, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Apfelsäure, meets USP/NF testing specifications
Sigma-Aldrich
DL-Apfelsäure, meets analytical specification of FCC, E296, 99-100.5% (alkalimetric)
Sigma-Aldrich
Natriumcarbonat-12C, 99.9 atom % 12C