Merck
Nanoscale analysis of ryanodine receptor clusters in dyadic couplings of rat cardiac myocytes.
Journal of molecular and cellular cardiology (2014-12-24)
Yufeng Hou, Isuru Jayasinghe, David J Crossman, David Baddeley, Christian Soeller
ZUSAMMENFASSUNG
The contractile properties of cardiac myocytes depend on the calcium (Ca(2+)) released by clusters of ryanodine receptors (RyRs) throughout the myoplasm. Accurate quantification of the spatial distribution of RyRs has previously been challenging due to the comparatively low resolution in optical microscopy. We have combined single-molecule localisation microscopy (SMLM) in a super-resolution modality known as dSTORM with immunofluorescence staining of tissue sections of rat ventricles to resolve a wide, near-exponential size distribution of RyR clusters that lined on average ~57% of the perimeter of each myofibril. The average size of internal couplons is ~63 RyRs (nearly 4 times larger than that of peripheral couplons) and the largest clusters contain many hundreds of RyRs. Similar to previous observations in peripheral couplons, we observe many clusters with one or few receptors; however ≥80% of the total RyRs were detected in clusters containing ≥100 receptors. ~56% of all clusters were within an edge-to-edge distance sufficiently close to co-activate via Ca(2+)-induced Ca(2+) release (100nm) and were grouped into 'superclusters'. The co-location of superclusters with the same or adjacent t-tubular connections in dual-colour super-resolution images suggested that member sub-clusters may be exposed to similar local luminal Ca(2+) levels. Dual-colour dSTORM revealed high co-localisation between the cardiac junctional protein junctophilin-2 (JPH2) and RyR clusters that confirmed that the majority of the RyR clusters observed are dyadic. The increased sensitivity of super-resolution images revealed approximately twice as many RyR clusters (2.2clusters/μm(3)) compared to previous confocal measurements. We show that, in general, the differences of previous confocal estimates are largely attributable to the limited spatial resolution of diffraction-limited imaging. The new data can be used to inform the construction of detailed mechanistic models of cardiac Ca(2+) signalling.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ACS reagent
Sigma-Aldrich
Pentan, anhydrous, ≥99%
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Glycerin, FCC, FG
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Saccharose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Saccharose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Saccharose, meets USP testing specifications