Merck
  • The N-N non-covalent domain of the nucleocapsid protein of type 2 porcine reproductive and respiratory syndrome virus enhances induction of IL-10 expression.
The N-N non-covalent domain of the nucleocapsid protein of type 2 porcine reproductive and respiratory syndrome virus enhances induction of IL-10 expression.
The Journal of general virology (2015-01-24)
Xing Liu, Baochao Fan, Juan Bai, Haiyan Wang, Yufeng Li, Ping Jiang
ZUSAMMENFASSUNG
Porcine reproductive and respiratory syndrome virus (PRRSV) usually establishes a prolonged infection and causes an immunosuppressive state. It has been proposed that IL-10 plays an important role in PRRSV-induced immunosuppression. However, this mechanism has not been completely elucidated. In this study, we found that transfection of 3D4/2 macrophages with the N protein gene of type 2 PRRSV significantly upregulated IL-10 expression at the transcriptional level. Moreover, alanine substitution mutation analysis revealed that the N protein residues 33-37, 65-68 and 112-123 were related to the upregulation of IL-10 promoter activity. Recombinant PRRSV with mutations at residues 33-37 in the N protein (rQ33-5A and rS36A) recovered from corresponding infectious cDNA clones and induced significantly lower levels of IL-10 production in infected monocyte-derived dendritic cells, as compared with their revertants rQ33-5A(R) and rS36A(R), and the wild-type recombinant PRRSV strain rNT/wt. These data indicate that the type 2 PRRSV N protein plays an important role in IL-10 induction and the N-N non-covalent domain is associated with this activity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Interleukin-4 human, IL-4, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
DL-Alanin, ≥99% (HPLC)
Sigma-Aldrich
IL-4 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
DL-Alanin, ≥99%, FCC, FG