Merck
  • Cloning, purification and evaluation of the enzymatic properties of a novel arylacetonitrilase from Luminiphilus syltensis NOR5-1B: a potential biocatalyst for the synthesis of mandelic acid and its derivatives.
Cloning, purification and evaluation of the enzymatic properties of a novel arylacetonitrilase from Luminiphilus syltensis NOR5-1B: a potential biocatalyst for the synthesis of mandelic acid and its derivatives.
Biotechnology letters (2015-04-10)
Huihui Sun, Wenyuan Gao, Haiyang Fan, Hualei Wang, Dongzhi Wei
ZUSAMMENFASSUNG
To examine nitrilase-mediated hydrolysis of nitriles to produce optically pure α-hydroxycarboxylic acids. A novel nitrilase, GPnor51, from Luminiphilus syltensis NOR5-1B was discovered by genomic data mining. It could hydrolyze racemic o-chloromandelonitrile to (R)-o-chloromandelic acid with high enantioselectivity (ee 98.2 %). GPnor51 was overexpressed in Escherichia coli BL21 (DE3), purified, and its catalytic properties studied. GPnor51 had a broad substrate acceptance toward various nitriles with structure diversity. It was an arylacetonitrilase that uses arylacetonitriles as optimal substrates. The V max and K m of GPnor51 towards o-chloromandelonitrile were 1.9 μmol min(-1) mg(-1) protein and 0.38 mM, respectively. GPnor51 also demonstrated high enantioselectivity toward mandelonitrile and other substituted mandelonitrile. This enzyme has a great potential for commercial production of optically pure (R)-mandelic acid and its derivatives.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Mandelsäure, 99%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.