Merck
  • An Oxofluoride Catalyst Comprised of Transition Metals and a Metalloid for Application in Water Oxidation.
An Oxofluoride Catalyst Comprised of Transition Metals and a Metalloid for Application in Water Oxidation.
Chemistry (Weinheim an der Bergstrasse, Germany) (2015-07-30)
Henrik Svengren, Shichao Hu, Ioannis Athanassiadis, Tanja M Laine, Mats Johnsson
ZUSAMMENFASSUNG
The application of the recently discovered oxofluoride solid solution (Cox Ni1-x )3 Sb4 O6 F6 as a catalyst for water oxidation is demonstrated. The phase exhibits a cubic arrangement of the active metal that forms oxo bridges to the metalloid with possible catalytic participation. The Co3 Sb4 O6 F6 compound proved to be capable of catalyzing 2H2 O→O2 +4H(+) +4e(-) at 0.33 V electrochemical and ≤0.39 V chemical overpotential with a TOF of 4.4⋅10(-3) , whereas Ni3 Sb4 O6 F6 needs a higher overpotential. Relatively large crystal cubes (0.3-0.5 mm) are easily synthesized and readily handled as they demonstrate both chemical resistance to wear after repeated in situ tests under experimental conditions, and have a mechanical hardness of 270 V0.1 using Vickers indentation. The combined properties of this compound offer a potential technical advantage for incorporation to a catalytic interface in future sustainable fuel production.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumphosphat, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Kaliumphosphat, ReagentPlus®
Sigma-Aldrich
Kaliumphosphat, Molecular Biology, ≥98.0%
Sigma-Aldrich
Kaliumphosphat, BioUltra, Molecular Biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Kaliumphosphat, 99.99% trace metals basis