Merck
  • A new plasmonic device made of gold nanoparticles and temperature responsive polymer brush on a silicon substrate.
A new plasmonic device made of gold nanoparticles and temperature responsive polymer brush on a silicon substrate.
Journal of colloid and interface science (2015-03-04)
Adem Zengin, Ugur Tamer, Tuncer Caykara
ZUSAMMENFASSUNG
This paper reports a general stepwise route assembling interface-mediated RAFT polymerization of 2-methoxyethoxy ethyl methacrylate and conversion of dodecyl trithiocarbonate end groups to thiol groups for gold nanoparticle assemblies. We intended by this way a new plasmonic device made of gold nanoparticles (Au NPs) and temperature responsive [poly((2-methoxyethoxy)ethyl) methacrylate] [poly(MEO2-MA)] brush on a silicon substrate. This polymeric layer replies to temperature changing by conformational variation and is therefore able to change the distance between the Au NPs on the brush layer with 5,5-dithiobis(2-dinitrobenzoic acid) (DTNB). We show that an increment of the external temperature reversibly stimulates a significant increase of the DTNB SERS signal.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
2,2′-Azobis(2-Methylpropionitril), 98%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.