Merck
  • Enzyme-triggered tyramine-enzyme repeats on prussian blue-gold hybrid nanostructures for highly sensitive electrochemical immunoassay of tissue polypeptide antigen.
Enzyme-triggered tyramine-enzyme repeats on prussian blue-gold hybrid nanostructures for highly sensitive electrochemical immunoassay of tissue polypeptide antigen.
Biosensors & bioelectronics (2015-06-13)
Tisen Xu, Haiying Zhang, Xuegui Li, Zhaohui Xie, Xiangyong Li
ZUSAMMENFASSUNG
A novel sandwich-type electrochemical immunoassay with sensitivity enhancement was developed for quantitative detection of tissue polypeptide antigen (TPA) by coupling with target-induced tyramine signal amplification on prussian blue-gold hybrid nanostructures. The immunosensor was prepared through immobilizing anti-TPA capture antibody on a cleaned screen-printed carbon electrode (SPCE). Prussian blue-gold hybrid nanostructures (PBGNS) labeled with horseradish peroxidase (HRP) and detection antibody were utilized as the signal-transduction tags. Upon target TPA introduction, the sandwiched immunocomplex was formed between capture antibody and detection antibody on the electrode. The carried HRP could trigger the formation of tyramine-HRP repeats on the PBGNS in the presence of H2O2. Using the doped prussian blue as the electron mediator, the conjugated HRP could catalyze the reduction of H2O2. Under the optimal conditions, the catalytic currents increased with the increasing target TPA in the dynamic range from 1.0 pg mL(-1) to 100 ng mL(-1) with a detection limit of 0.3 pg mL(-1). The reproducibility and specificity of the electrochemical immunoassay were acceptable. In addition, the contents of target TPA in nine human serum specimens were evaluated by using the developed electrochemical immunosensor, and the obtained results correlated well with those from commercially enzyme-linked immunosorbent assay (ELISA) method with a correlation coefficient of 0.9975.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Hydroxysuccinimid, 98%
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
1,2-Dichlorethan, anhydrous, 99.8%
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyd -Lösung, 50 wt. % in H2O
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrazin Monohydrat, N2H4 64-65 %, reagent grade, ≥97%
Sigma-Aldrich
Hydrazin Hydrat, reagent grade, N2H4 50-60 %
Sigma-Aldrich
Salzsäure -Lösung, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
Tyramin, ≥98.0%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Salzsäure -Lösung, 32 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutarsäure-Dialdehyd -Lösung, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyd -Lösung, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Tyramin, 98%, FG