Merck
Resistance exercise induces a greater irisin response than endurance exercise.
Metabolism: clinical and experimental (2015-06-18)
Yoshifumi Tsuchiya, Daisuke Ando, Kaoru Takamatsu, Kazushige Goto
ZUSAMMENFASSUNG
We determined detailed time-course changes in the irisin response to acute exercise using different exercise modes. In experiment 1, seven healthy males rested for 12h (8:00-20:00) to determine the diurnal variation in plasma irisin concentration. In experiment 2, 10 healthy males conducted three exercises to clarify time-course changes in plasma irisin concentration over 6h, using a randomized crossover design. The resistance exercise (R) trial consisted of eight exercises of 12 repetitions with 3-4 sets at 65% of one repetition maximum (1RM). The endurance exercise (E) trial consisted of 60 min of pedaling at 65% of maximal oxygen uptake (VO2max). In the combined mode (R+E) trial, 30 min of endurance exercise was preceded by 30 min of resistance exercise. In experiment 1, no significant changes in plasma irisin concentration were observed over 12h. In experiment 2, the R trial showed a marked increase in plasma irisin concentration 1h after exercise (P<0.05), but not in the E or R+E trials. The area under the curve (AUC) for irisin concentrations for 6h after exercise was significantly higher in the R trial than in the R+E trial (P<0.05). The AUC for irisin concentrations was significantly correlated with AUC values for blood glucose, lactate, and serum glycerol (r=0.37, 0.45, 0.45, respectively. P<0.05). Resistance exercise resulted in significantly greater irisin responses compared with endurance exercise alone, and resistance and endurance exercises combined.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerin, FCC, FG
Sigma-Aldrich
Glycerin, meets USP testing specifications